C89803
4-Cyanobenzoic acid
99%
Synonym(s):
Terephthalic acid mononitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NCC6H4CO2H
CAS Number:
Molecular Weight:
147.13
Beilstein:
1862865
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99%
form
liquid crystal
mp
219-221 °C (dec.) (lit.)
SMILES string
OC(=O)c1ccc(cc1)C#N
InChI
1S/C8H5NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,(H,10,11)
InChI key
ADCUEPOHPCPMCE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Daniel L Kirschner et al.
Analytical chemistry, 79(2), 736-743 (2007-01-16)
A capillary electrophoresis method with laser-induced fluorescence detection for the chiral separation of cyanobenz[f]isoindole (CBI) derivatives of amino acids was developed and optimized. The enantioseparations are accomplished with sulfated beta-CD (S-beta-CD) as chiral selector at low pH and reverse polarity.
James Turkson et al.
Molecular cancer therapeutics, 3(3), 261-269 (2004-03-18)
The critical role of signal transducer and activator of transcription 3 (Stat3) in the growth and survival of human tumor cells identifies it as a promising target for cancer drug discovery. We previously identified a Stat3 SH2 domain-binding phosphopeptide, PY*LKTK
Federico Polo et al.
Journal of the American Chemical Society, 127(2), 492-493 (2005-01-13)
The rate constant of intramolecular electron transfer through oligopeptides based on the alpha-aminoisobutyric acid residue was determined as a function of the peptide length and found to depend weakly on the donor-acceptor separation. By measuring the electron-transfer activation energy and
P Boudeville et al.
Journal of pharmaceutical sciences, 84(9), 1083-1089 (1995-09-01)
A new methodology of pH-metric data treatment was developed to extract the stoichiometry and the thermodynamic association constants of guest-cyclodextrin inclusion complexes in dilute aqueous solution when the guest is a participant in an acid-base equilibrium. pH-metric titration curves in
Lisa A Hoferkamp et al.
Environmental science & technology, 40(7), 2206-2212 (2006-05-02)
The reductive transformation of p-cyanonitrobenzene (pCNB) was investigated in laboratory batch slurries exhibiting dominant terminal electron accepting processes (TEAPs). Pseudo-first-order rate constants (k(obs)) were measured for the reduction of pCNB in nitrate-reducing, iron-reducing, sulfate-reducing, and methanogenic sediment slurries. Reduction was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service