Skip to Content
Merck
All Photos(2)

Key Documents

I10201

Sigma-Aldrich

4-Iodophenol

99%

Synonym(s):

1-Iodo-4-hydroxybenzene, 4-Hydroxyiodobenzene, 4-Hydroxyphenyl iodide, p-Hydroxyiodobenzene, p-Iodophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4OH
CAS Number:
Molecular Weight:
220.01
Beilstein:
1904544
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

138 °C/5 mmHg (lit.)

mp

92-94 °C (lit.)

SMILES string

Oc1ccc(I)cc1

InChI

1S/C6H5IO/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

VSMDINRNYYEDRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Iodophenol can be used as a building block for the synthesis of:
  • Hydroxybiaryls by reacting with aryl boronic acids via Pd-catalyzed Suzuki-Miyaura coupling reaction.
  • Aryl substituted olefins by reacting with acrylates via Pd-catalyzed Heck reaction.
  • Iodinated-4-aryloxymethylcoumarins
  • Hydroxylated stilbenoids and psammaplysenes A and B.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure-activity relationship of iodinated-4-aryloxymethyl-coumarins as potential anti-cancer and anti-mycobacterial agents
Basanagouda, M, et al.
European Journal of Medicinal Chemistry, 74(1), 225-233 (2014)
Carbon- carbon coupling reactions catalyzed by heterogeneous palladium catalysts
Yin L and Liebscher J
Chemical Reviews, 107(1), 133-173 (2007)
Pd/C as a reusable catalyst for the coupling reaction of halophenols and arylboronic acids in aqueous media
Sakurai H, et al.
The Journal of Organic Chemistry, 67(8), 2721-2722 (2002)
Total synthesis of psammaplysenes A and B, naturally occurring inhibitors of FOXO1a nuclear export
Georgiades SN and Clardy J
Organic Letters, 7(19), 4091-4094 (2005)
Haigang Li et al.
The Analyst, 134(4), 800-804 (2009-03-24)
Simple and sensitive chemiluminescence (CL) and CL imaging methods have been developed for the magnetic bead-based DNA hybridization assay. The assay relies on the high sensitivity and long stable light signal of the CL system in which horseradish peroxidase (HRP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service