Skip to Content
Merck
All Photos(2)

Documents

C83201

Sigma-Aldrich

β-Citronellol

95%

Synonym(s):

(±)-β-Citronellol, beta-Citronellol, 3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
Beilstein:
1721507
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.4 (vs air)

vapor pressure

~0.02 mmHg ( 25 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

222 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

SMILES string

CC(CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3

InChI key

QMVPMAAFGQKVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renan G Brito et al.
Basic & clinical pharmacology & toxicology, 112(4), 215-221 (2012-10-06)
Citronellol (CT) is a monoterpenoid alcohol present in the essential oil of many medicinal plants, such as Cymbopogon citratus. We evaluated the antinociceptive effects of CT on orofacial nociception in mice and investigated the central pathway involved in the effect.
M Spiller et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 120(7), 1461-1471 (2010-01-20)
The scent of flowers is a very important trait in ornamental roses in terms of both quantity and quality. In cut roses, scented varieties are a rare exception. Although metabolic profiling has identified more than 500 scent volatiles from rose
Karin Förster-Fromme et al.
Applied microbiology and biotechnology, 87(3), 859-869 (2010-05-22)
Terpenes are a huge group of natural compounds characterised by their predominantly pleasant smell. They are built up by isoprene units in cyclic or acyclic form and can be functionalised by carbonyl, hydroxyl or carboxyl groups and by presence of
Yoshikazu Kitano et al.
Biofouling, 27(2), 201-205 (2011-02-01)
Twenty novel simple alkyl isocyanides derived from citronellol were synthesized and evaluated for their antifouling activity and toxicity against cypris larvae of the barnacle, Balanus amphitrite. The anti-barnacle activity of the synthesized isocyanides was in the EC(50) range of 0.08-1.49
Michiko Katsukawa et al.
Bioscience, biotechnology, and biochemistry, 75(5), 1010-1012 (2011-05-21)
We evaluated the effects of rose oil on the peroxisome proliferator-activated receptor (PPAR) and cyclooxygenase-2 (COX-2). Citronellol and geraniol, the major components of rose oil, activated PPARα and γ, and suppressed LPS-induced COX-2 expression in cell culture assays, although the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service