Skip to Content
Merck
All Photos(1)

Key Documents

303461

Sigma-Aldrich

(R)-(+)-β-Citronellol

95%

Synonym(s):

(+)-β-Citronellol, (R)-3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
Beilstein:
1721506
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

~0.02 mmHg ( 25 °C)

Quality Level

Assay

95%

form

liquid

optical activity

[α]19/D +5.3°, neat

refractive index

n20/D 1.456 (lit.)

bp

112-113 °C/12 mmHg (lit.)

density

0.857 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@@H](CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m1/s1

InChI key

QMVPMAAFGQKVCJ-SNVBAGLBSA-N

Application

(R)-(+)-β-Citronellol can be used to prepare:
  • (R)-(+)-β-citronellyl acetate by treating with vinyl acetate via trans esterification reaction using immobilized Rhizomucor miehei lipase as biocatalyst.[1]
  • (R)-(+)-citronellal and (R)-(+)-citronellic acid by the oxidation reaction.[2]
  • Citronellol aziridine by treating with 2,2,2-trichloroethoxysulfonamide by intermolecular olefin aziridination.[3]

It can also be used as a starting material for the synthesis of (+)-integerrinecic acid lactone.[4]
(R)-(+)-β-Citronellol can undergo:
  • Oxidation to form (R)-(+)-citronellal,[5] an antifungal agent[6] and (R)-(+)-citronellic acid.[7]
  • Intermolecular olefin aziridination with 2,2,2-trichloroethoxysulfonamide to form citronellol aziridine for ring-opening reactions.[3]
  • Series of reactions to form α2δ-ligands for treating generalized anxiety disorder and insomnia.[8]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel synthesis of (+)-integerrinecic acid lactone from R-(+) β-citronellol
White JD and Jayasinghe LR
Tetrahedron Letters, 29(18), 2139-2142 (1988)
Rh-catalyzed alkene oxidation: a highly efficient and selective process for preparing N-alkoxysulfonyl aziridines.
Guthikonda K, et al.
Tetrahedron, 62(49), 11331-11342 (2006)
Phenylglycine methyl ester, a useful tool for absolute configuration determination of various chiral carboxylic acids.
Yabuuchi T and Kusumi T
The Journal of Organic Chemistry, 65(2), 397-404 (2000)
Abir B Majumder et al.
Chemistry Central journal, 1, 10-10 (2007-09-21)
Use of enzymes in low water media is now widely used for synthesis and kinetic resolution of organic compounds. The frequently used enzyme form is the freeze-dried powders. It has been shown earlier that removal of water molecules from enzyme
A very mild and chemoselective oxidation of alcohols to carbonyl compounds.
De Luca L, et al.
Organic Letters, 3(19), 3041-3043 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service