Skip to Content
Merck
All Photos(3)

Key Documents

B4007

Sigma-Aldrich

1,2,4,5-Benzenetetracarboxylic acid

96%

Synonym(s):

Pyromellitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2(CO2H)4
CAS Number:
Molecular Weight:
254.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

mp

281-284.5 °C (lit.)

SMILES string

OC(=O)c1cc(C(O)=O)c(cc1C(O)=O)C(O)=O

InChI

1S/C10H6O8/c11-7(12)3-1-4(8(13)14)6(10(17)18)2-5(3)9(15)16/h1-2H,(H,11,12)(H,13,14)(H,15,16)(H,17,18)

InChI key

CYIDZMCFTVVTJO-UHFFFAOYSA-N

Related Categories

Application

1,2,4,5-Benzenetetracarboxylic acid is extensively used as a ligand in the synthesis of a wide range of metal-organic frameworks (MOFs) and coordination polymers. It can also be used to prepare supramolecular hydrogels by synthesizing gelators reacting 1,2,4,5-benzenetetracarboxylic acid with 4-hydroxy pyridine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

617.0 °F

Flash Point(C)

325 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid Barkleit et al.
Inorganic chemistry, 50(12), 5451-5459 (2011-05-25)
Thermodynamic parameters for the complexation of Eu(3+) with pyromellitic acid (1,2,4,5-benzenetetracarboxylic acid, BTC) as a model system for polymerizable metal-complexing humic acids were determined using temperature-dependent time-resolved laser-induced fluorescence spectroscopy (TRLFS) and isothermal titration calorimetry (ITC). At low metal and
X Xiong et al.
Electrophoresis, 19(12), 2243-2251 (1998-10-07)
The simultaneous separation and detection of small cations and anions by capillary zone electrophoresis (CZE) with indirect ultraviolet (UV) detection was successfully demonstrated in a background electrolyte (BGE) containing two UV-absorbing components. Benzylamine, imidazole, benzenesulfonic acid, sulfosalicylic acid, and pyromellitic
Jacek Gawroński et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 8(11), 2484-2494 (2002-08-16)
The chiral but highly symmetrical acyclic and cyclic pyromellitic diimide dimers and trimers 2-5 have been obtained and characterized for the first time. The pyromellitdiimide chromophores in these molecules are linked by a rigid diequatorially 1,2-disubstituted cyclohexane skeleton. The structures
Yu Liu et al.
Carbohydrate research, 338(17), 1751-1757 (2003-08-02)
A novel bridged bis(beta-cyclodextrin) with a pyromellitic acid 2,5-diamide tether (2) has been synthesized by reaction of 6(I)-(2-aminoethyleneamino)-6-deoxycyclomaltoheptaose [mono 6-(2-aminoethyleneamino)-6-deoxy-beta-cyclodextrin] with 1,2,4,5-benzenetetracarboxylic dianhydride. Its inclusion complexation behavior with some representative dyestuffs, i.e., Acridine Red (AR), Rhodamine B (RhB), Neutral Red
1, 2, 4, 5?Benzenetetracarboxylic Acid and 4, 4??Bipyridine as Ligands in Designing Low?Dimensional Coordination Polymers.
Ruiz?Perez C, et al.
European Journal of Inorganic Chemistry, 2004(19), 3873-3879 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service