75617
2-Deoxy-L-ribose
≥97.0% (TLC)
Synonym(s):
2-Deoxy-L-erythro-pentose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥97.0% (TLC)
form
powder
optical activity
[α]/D 54.0±2.0°, 24 hr, c = 1 in H2O
storage temp.
2-8°C
SMILES string
O=CC[C@@H](O)[C@@H](O)CO
InChI
1S/C5H10O4/c6-2-1-4(8)5(9)3-7/h2,4-5,7-9H,1,3H2/t4-,5+/m1/s1
InChI key
ASJSAQIRZKANQN-UHNVWZDZSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Inhibitor of enzymes and valuable building block for synthesis
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Masao Shiozaki et al.
Carbohydrate research, 370, 46-66 (2013-03-05)
KRN7000 is one of the α-galactosylceramides, which has a 2-hexacosanoylamino-3,4-dihydroxyoctadecyl group. This compound, known as a ligand for the activation of CD1d mediated invariant natural killer T cells (iNKT cells) which release both T helper 1 (Th1) cytokines such as
Aqeel Ahmed et al.
Journal of the American Chemical Society, 128(44), 14224-14225 (2006-11-02)
The reaction of 70 unprotected, diversely functionalized free reducing sugars with methoxyamine-appended colchicine led to the production of a 58-member glycorandomized library. High-throughput cytotoxicity assays revealed glycosylation to modulate specificity and potency. Library members were also identified which, unlike the
Ryuji Ikeda et al.
Biochemical and biophysical research communications, 291(4), 806-812 (2002-02-28)
An angiogenic factor, platelet-derived endothelial cell growth factor/thymidine phosphorylase (PD-ECGF/TP), stimulates the chemotaxis of endothelial cells and confers resistance to apoptosis induced by hypoxia. 2-deoxy-D-ribose, a degradation product of thymidine generated by TP enzymatic activity partially prevented hypoxia-induced apoptosis. 2-Deoxy-D-ribose
Warfarin glycosylation invokes a switch from anticoagulant to anticancer activity.
Pauline Peltier-Pain et al.
ChemMedChem, 6(8), 1347-1350 (2011-06-30)
Yuichi Nakajima et al.
Cancer research, 64(5), 1794-1801 (2004-03-05)
Thymidine phosphorylase (TP) catalyzes the reversible conversion of thymidine to thymine, thereby generating 2-deoxy-D-ribose-1-phosphate, which upon dephosphorylation forms 2-deoxy-D-ribose (D-dRib), a degradation product of thymidine. We have previously shown that D-dRib promotes angiogenesis and chemotaxis of endothelial cells and also
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service