Skip to Content
Merck
All Photos(1)

Key Documents

A8784

Sigma-Aldrich

N-Acetylputrescine hydrochloride

≥98% (TLC)

Synonym(s):

Monoacetyl-1,4-diaminobutane hydrochloride, N-(4-Aminobutyl)acetamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O · HCl
CAS Number:
Molecular Weight:
166.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (TLC)

form

solid

storage temp.

2-8°C

SMILES string

Cl.CC(=O)NCCCCN

InChI

1S/C6H14N2O.ClH/c1-6(9)8-5-3-2-4-7;/h2-5,7H2,1H3,(H,8,9);1H

InChI key

XBECFEJUQZXMFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W J Hrushesky et al.
Cancer research, 43(8), 3944-3947 (1983-08-01)
Polyamines are small, highly charged, organic cations of possible regulatory importance in RNA-dependent protein synthesis, the production of which reflects cellular growth and division. The cytokinetics of normal cell populations is circadian rhythmic. This is reflected by a circadian rhythmicity
S Yamamoto et al.
Journal of chromatography, 233, 29-38 (1982-12-10)
A gas chromatographic method was developed for the determination of monoacetylputrescine, monoacetylcadaverine, N1-acetylspermidine and N8-acetylspermidine in human urine. The amines were isolated from urine by silica gel column chromatography. 1,10-Diaminodecane was used as internal standard. The amines were reacted with
R M Wittich et al.
Molecular and biochemical parasitology, 38(1), 13-17 (1990-01-01)
A novel type of N-acetyltransferase, clearly different from the nuclear and cytosolic polyamine N-acetyltransferases of mammals, was recently found in the intestinal nematode Ascaris suum. The occurrence of this putrescine N-acetylating enzyme has also been noted in the filarial parasite
J Lacy et al.
Journal of immunology (Baltimore, Md. : 1950), 135(6), 3772-3776 (1985-12-01)
N,N'-Diacetylputrescine (tetramethylenebisacetamide [TMBA]) and its six carbon analog, hexamethylenebisacetamide (HMBA), inhibited the proliferative response of human B lymphocytes to anti-mu and formalinized Cowan I strain Staphylococcal aureus (SAC) stimulation. In contrast, B cell growth factor-stimulated proliferation of human B cells
P Kishore et al.
Microbios, 73(294), 7-21 (1993-01-01)
1,3-Diaminopropane has been identified as the major polyamine of Acanthamoeba culbertsoni. N-acetylputrescine and spermidine were present in appreciable amounts and putrescine as well as N-acetylspermidine were also detected, but spermine was absent. Changes in polyamine levels were observed during the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service