Skip to Content
Merck
All Photos(1)

Key Documents

A21401

Sigma-Aldrich

4-Acetylpyridine

97%

Synonym(s):

Methyl 4-pyridyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
Beilstein:
107629
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

212 °C (lit.)

mp

13-16 °C (lit.)

density

1.095 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccncc1

InChI

1S/C7H7NO/c1-6(9)7-2-4-8-5-3-7/h2-5H,1H3

InChI key

WMQUKDQWMMOHSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Imamura et al.
Journal of biochemistry, 125(1), 41-47 (1999-01-09)
An enzyme responsible for the ketone-reduction of 4-benzoylpyridine (4BP) was purified 350-fold to homogeneity from the cytosolic fraction of rabbit heart. The purified enzyme exhibited a molecular mass of 110 kDa on gel filtration, and 27 kDa on SDS-PAGE, indicating
Y Imamura et al.
Biochemistry and molecular biology international, 33(5), 893-899 (1994-08-01)
A correlation was observed between the values of specificity constant (kcat/Km) of carbonyl reductase from rabbit liver for acetohexamide analogs and their partition coefficients. This result indicates that the hydrophobicity in straight-chain alkyl groups of acetohexamide analogs plays an important
A Hall et al.
Neuroscience letters, 85(1), 110-112 (1988-02-15)
4-Acetylpyridine, earlier reported by us to be an anticonvulsant, offers long-lasting protection after a single administration against hypothermic restraint stress-induced gastric ulceration in mice. Electroshock convulsions, marginally but not significantly protective against such ulcers themselves, when coupled with 4-acetylpyridine administration
S G Zhu et al.
Brain research, 481(2), 356-360 (1989-03-06)
Measurements of striatal choline acetyltransferase (ChAT) and glutamic acid decarboxylase (GAD) activities indicated that systemic administration of 4-8 mg/kg of MK-801 to rats completely blocked neuronal damage due to intrastriatal injections of 75-150 nmol of quinolinic acid. Similar experiments with
Y Imamura et al.
Biochemistry and molecular biology international, 31(6), 1105-1110 (1993-12-01)
Carbonyl reductase from rabbit kidney was rapidly inactivated by diethylpyrocarbonate (DEPC). A similar inactivation was observed in photooxidation of the enzyme by methylene blue. The inactivation by DEPC was time- and concentration-dependent and followed pseudo-first-order kinetics. The results obtained from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service