Skip to Content
Merck
All Photos(1)

Key Documents

935166

Sigma-Aldrich

Hyaluronic acid

acid form, average Mw 25,000

Synonym(s):

HA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C16H27NO11)n
CAS Number:
MDL number:
UNSPSC Code:
51202002
NACRES:
NA.21

description

acid form of hylauronic acid

Quality Level

form

powder or chunks (or Fibers)

mol wt

average Mw 25,000

color

white to faint yellow, Faint Brown

application(s)

advanced drug delivery
(Tissue engineering)

storage temp.

2-8°C

SMILES string

N(C1C(OC(C(C1OC4OC(C(C(C4O)O)O)C(=O)O)O)CO)OC2C(OC(C(C2O)O)OC3C(C(OC(C3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45)

InChI key

KIUKXJAPPMFGSW-UHFFFAOYSA-N

General description

Hyaluronic acid (HA) is a a natural polymer found in many extracellular matrices (ECM). Its biocompatibility, biodegradability, and biomimetic chacteristics make it one of the most advantagous biopolymers for myriad of biomedical applications including drug delivery, tissue engineering, hydrogel development, cosmetics, regenerative medicine and 3D bioprinting.
Unlike other HA products which are typically available in a sodium salt form, HA acid form is soluble in organic solvents such as DMSO. This allows for a larger range of possible chemical modifications and functionalization. For instance, the carboxylic acid of the glucoronic acid can be modified or functionalized with hydrazine end groups.

Application

  • Tissue engineering including cartilage tissue engineering, bone tissue engineering, and other musculoskeletal tissues
  • Drug delivery and a carrier of small molecules, large molecules, and biomolecules
  • Biosensors, films and transdermal microneedles development
  • Medical device coatings, wound healing, and cosmetics

Features and Benefits

  • The same advantages of HA but in acid form
  • Soluble in organic solvents such as DMSO
  • Larger range of chemical modifications and functionalization possibilities

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyaluronic Acid: Molecular Mechanisms and Therapeutic Trajectory
Gupta R C, et al.
Frontiers in veterinary science, 25, 192-192 (2019)
Thiolated Hyaluronic Acid as Versatile Mucoadhesive Polymer: From the Chemistry Behind to Product Developments-What Are the Capabilities?
Griesser J, et al.
Polymers (Basel, Switzerland), 28, 243-243 (2018)
A mild and straightforward one-pot hyaluronic acid functionalization through termination of poly-(2-alkyl-2-oxazoline)
Madau M, et al.
Polymer, 230, 124059-124059 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service