Skip to Content
Merck
All Photos(4)

Documents

H5388

Sigma-Aldrich

Hyaluronic acid sodium salt from rooster comb

avian glycosaminoglycan polysaccharide

Synonym(s):

Poly(β-glucuronic acid-[1→3]-β-N-acetylglucosamine-[1→4]), alternating

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

chicken (rooster comb)

Quality Level

form

powder

impurities

≤2% protein (Lowry)

color

white

solubility

water: ~5 g/L

cation traces

Na: 3-7%

storage temp.

−20°C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Application

Hyaluronic acid sodium salt from rooster comb has been used:
  • as a component of bi-layer gelatin scaffold
  • as a vehicle for release of gentamicin during left ear surgery in guinea pigs
  • in hyaluronic acid binding assay in human embryonic kidney cell line 293T

Biochem/physiol Actions

High molecular mass polymer composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine which forms the core of complex proteoglycan aggregates found in extracellular matrix.
Hyaluronic acid is a polysaccharide present in the skin, connective tissue, vitreous humor and synovial fluid. It is essential for tissue hydration and functions to regenerate skin post injury or wounds. Hyaluronic acid, apart from having dermatologic and ophthalmologic applications, serves as a drug delivery vehicle. Hyaluronic acid also exhibits antioxidant and antiglycation properties.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and characterization of hyaluronic acid from chicken combs
Rosa CS, et al.
Ciencia Rural, 42(9), 1682-1687 (2012)
Hyaluronan binding motifs of USP17 and SDS3 exhibit anti-tumor activity
Ramakrishna S, et al.
PLoS ONE, 7(5), e37772-e37772 (2012)
Extraction, purification and characterization of hyaluronic acid from Rooster comb
Kulkarni SS, et al.
Journal of Applied and Natural Science, 10(1), 313-315 (2018)
Ya-li Yang et al.
Biophysical journal, 96(4), 1566-1585 (2009-02-17)
In this work, the gelation of three-dimensional collagen and collagen/hyaluronan (HA) composites is studied by time sweep rheology and time lapse confocal reflectance microscopy (CRM). To investigate the complementary nature of these techniques, first collagen gel formation is investigated at
Hyaluronic acid enhances gene delivery into the cochlea
Shibata SB, et al.
Human Gene Therapy, 23(3), 302-310 (2011)

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

See All

Protocols

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service