Skip to Content
Merck
All Photos(1)

Documents

924490

Sigma-Aldrich

Hyaluronic acid methacrylate

Low Viscosity, Low Endotoxin, 0.2 um sterile filtered, 0.2 μm, sterile-filtered

Synonym(s):

HAMA, Hyaluronan, Hyaluronic acid, MeHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NaC20H28NO15)n
UNSPSC Code:
12352106
NACRES:
NA.23

Quality Level

description

Validation : HNMR at 40°C

sterility

sterile-filtered

form

(Solid chunks, fibers or powder)

impurities

≤10 CFU/g Bioburden (Fungal)
≤10 CFU/g Bioburden (Total Aerobic)
100 EU/g Endotoxin

color

white to pale yellow

particle size

0.2 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Hyaluronic acid (HA) is a linear polysaccharide of alternating D-glucuronic acid and N-acetyl-D-glucosamine found primarily in connective tissues. HA based hydrogels are widely used in tissue engineering, 3D bioprinting, and drug deliery applications. The methacrylate functionalized hyaluronic acid is photo-crosslinkable, and can be used to generate crosslinked hydrogels.

Packaging

1EA = 500 mg of lyophilized material in glass bottle.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stiffness and adhesivity control aortic valve interstitial cell behavior within hyaluronic acid based hydrogels.
Duan, et al.
Acta Biomaterialia, 9, 7640-7650 (2018)
Photocrosslinkable Hyaluronan-Gelatin Hydrogels for Two-Step Bioprinting
Skardal, et al.
Tissue Engineering: Part A, 16 (8) (2010)
Micromolding of shape-controlled, harvestable cell-laden hydrogels.
Yeh, et al.
Biomaterials, 27, 5391-5398 (2007)
Influence of Three-Dimensional Hyaluronic Acid Microenvironments on Mesenchymal Stem Cell Chondrogenesis
Chung and Burdick
Tissue Engineering: Part A, 15(2) (2009)
Judy Yeh et al.
Biomaterials, 27(31), 5391-5398 (2006-07-11)
Encapsulation of mammalian cells within hydrogels has great utility for a variety of applications ranging from tissue engineering to cell-based assays. In this work, we present a technique to encapsulate live cells in three-dimensional (3D) microscale hydrogels (microgels) of controlled

Articles

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service