Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hyaluronic acid methacrylate
Linear Formula:
(NaC
20
H
28
NO
15
)n
Products
Brand
Sigma-Aldrich (4)
Color
Physical Form
Quality Segment
Particle Size (µm)
Polymer Composition
Polymer Type
Sterilization Method
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
914800
Hyaluronic acid methacrylate,
Degree of substitution: 10% - 30%, M
w
170,000-250,000
Expand
View Pricing
914568
Hyaluronic acid methacrylate,
average degree of substitution 35%, average M
w
55000
Expand
View Pricing
914304
Hyaluronic acid methacrylate,
average degree of substitution 35%, average M
w
175000
Expand
View Pricing
924490
Hyaluronic acid methacrylate,
Low Viscosity, Low Endotoxin, 0.2 um sterile filtered, 0.2 μm, sterile-filtered
Expand
View Pricing