Skip to Content
Merck
All Photos(1)

Key Documents

917567

Sigma-Aldrich

Indium tin oxide coated PEN with Pt layer

60% transparency, size 9.6 cm × 9.6 cm × 125 μm, sheet resistance 5 Ω/sq

Synonym(s):

ITO coated polyethylene naphthalate with Pt layer, ITO-PEN with Pt layer (60% transparency), PECF-CAT03

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

Quality Level

description

Catalyst material: Pt/Ti alloy
Substrate: polyethylene naphthalate (PEN) film 125 micron thick (one side coated)

form

solid

sheet resistance

5 Ω/sq

size

9.6 cm × 9.6 cm × 125 μm

SMILES string

C

InChI

1S/CH4/h1H4

InChI key

VNWKTOKETHGBQD-UHFFFAOYSA-N

Application

This product is a one-side coated, flexible, transparent, ready-to-use, low sheet resistance, ITO coated PEN polymer film. Pt catalyste was sputtered onto the coated side with 60% final transmittance.
This product can be used for rapid fabrication of flexible Perovskite solar cells, sensors and dye-sensitized solar cells.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photovoltaic Performance of Plastic Dye-Sensitized Electrodes Prepared by Low-Temperature Binder-Free Coating of Mesoscopic Titania.
Miyasaka T, et al.
Journal of the Electrochemical Society (2007)
Jennifer M Pringle et al.
Chemical communications (Cambridge, England), 46(29), 5367-5369 (2010-06-23)
Electrodeposition of poly(3,4-ethylenedioxythiophene) onto conducting plastic allows the cheap and facile synthesis of plastic cathodes for dye-sensitised solar cells, having excellent solar cell efficiency (8%), at a fraction of the cost of platinised plastic.
Low temperature fabrication of high performance and transparent Pt counter electrodes for use in flexible dye-sensitized solar cells.
Xiao Y, et al.
Science Bulletin, 57, 2329-2334 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service