- Electrodeposited PEDOT-on-plastic cathodes for dye-sensitized solar cells.
Electrodeposited PEDOT-on-plastic cathodes for dye-sensitized solar cells.
Chemical communications (Cambridge, England) (2010-06-23)
Jennifer M Pringle, Vanessa Armel, Douglas R MacFarlane
PMID20567792
ABSTRACT
Electrodeposition of poly(3,4-ethylenedioxythiophene) onto conducting plastic allows the cheap and facile synthesis of plastic cathodes for dye-sensitised solar cells, having excellent solar cell efficiency (8%), at a fraction of the cost of platinised plastic.
Sigma-Aldrich
Indium tin oxide coated PEN, with 0% transparent Pt, L × W × thickness 9.6 cm × 9.6 cm × 125 μm, sheet resistance 5 Ω/sq
Sigma-Aldrich
Indium tin oxide coated PEN with Pt layer, 60% transparency, size 9.6 cm × 9.6 cm × 125 μm, sheet resistance 5 Ω/sq