Skip to Content
Merck
All Photos(1)

Key Documents

86720

Sigma-Aldrich

Tetraethylthiuram disulfide

≥97.0% (S)

Synonym(s):

Bis(diethylthiocarbamoyl) disulfide, Bis(diethylthiocarbamyl) disulfide, Disulfiram, TETD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSS2CSN(C2H5)2
CAS Number:
Molecular Weight:
296.54
Beilstein:
1712560
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (S)

form

powder

mp

69-71 °C (lit.)

functional group

amine
disulfide

SMILES string

CCN(CC)C(=S)SSC(=S)N(CC)CC

InChI

1S/C10H20N2S4/c1-5-11(6-2)9(13)15-16-10(14)12(7-3)8-4/h5-8H2,1-4H3

InChI key

AUZONCFQVSMFAP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Alcohol dehydrogenase inhibitor.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter M Meggyesy et al.
Molecules (Basel, Switzerland), 25(21) (2020-10-31)
The therapeutic utility of the copper ionophore disulfiram was investigated in a diet-induced obesity mouse model (C57BL/6J background), both through administration in feed (0.05 to 1% (w/w)) and via oral gavage (150 mg/kg) for up to eight weeks. Mice were
Beatriz Mothe et al.
The Journal of antimicrobial chemotherapy, 70(6), 1833-1842 (2015-03-01)
The safety, immunogenicity, impact on the latent reservoir and rebound of viral load after therapeutic HIV-1 vaccination with recombinant modified vaccinia Ankara-based (MVA-B) HIV-1 vaccine expressing monomeric gp120 and the fused Gag-Pol-Nef polyprotein of clade B with or without a
Charlotte H Jørgensen et al.
Alcoholism, clinical and experimental research, 35(10), 1749-1758 (2011-05-28)
Alcohol use disorders (AUD) involving hazardous, harmful, and addictive misuse of alcohol are widespread in most parts of the world. The aim of this study was to review the effect of disulfiram in the treatment of patients with AUD. The
Meriem Gaval-Cruz et al.
Molecular interventions, 9(4), 175-187 (2009-09-02)
The anti-alcoholism drug disulfiram (Antabuse), which is an inhibitor of aldehyde dehydrogenase, induces an aversive reaction to alcohol consumption and thereby helps patients reduce alcohol intake. Recent clinical trials, initiated to investigate whether disulfiram could be used to treat individuals
Boris Cvek et al.
Drug discovery today, 13(15-16), 716-722 (2008-06-27)
The major approach to the development of anticancer drugs involves searching for new compounds, efficient against malignancies, which are not, as yet, used clinically. This strategy is time-consuming and expensive. Recent studies have disclosed a surprising, but mechanistically consistent, anticancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service