Skip to Content
Merck
All Photos(1)

Key Documents

253901

Sigma-Aldrich

5-Bromo-1-pentene

95%

Synonym(s):

1-Bromo-4-pentene, 4-Pentenyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3CH=CH2
CAS Number:
Molecular Weight:
149.03
Beilstein:
506077
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.463 (lit.)

bp

126-127 °C/765 mmHg (lit.)

density

1.258 g/mL at 25 °C (lit.)

functional group

alkyl halide
allyl
bromo

storage temp.

2-8°C

SMILES string

BrCCCC=C

InChI

1S/C5H9Br/c1-2-3-4-5-6/h2H,1,3-5H2

InChI key

LPNANKDXVBMDKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-1-pentene was used in stereoselective synthesis of 7α-(3-carboxypropyl) estradiol. It was used in preparation of thioacetate 11 of sialic acid having thioglycosidic linkage. It was also used as staring material in recent syntheses of DL-histrionicotoxin and benzophenone-containing fatty acids.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maheswaran S Karatholuvhu et al.
Journal of the American Chemical Society, 128(39), 12656-12657 (2006-09-28)
The synthesis of (+/-)-histrionicotoxin has been achieved in just nine steps using a two-directional synthesis strategy. Key reactions include a two-directional cross-metathesis, a tandem oxime formation/Michael addition/1,4-prototopic shift/[3 + 2]-cycloaddition cascade, a selective Z,Z-bisenyne formation, and a one-pot N-O and
Jun-Ichi Sakamoto et al.
Bioorganic & medicinal chemistry letters, 17(3), 717-721 (2006-11-11)
An efficient synthesis of a series of carbosilane dendrimers uniformly functionalized with alpha-thioglycoside of sialic acid was accomplished. The results of a preliminary study on biological responses against influenza virus sialidases using thiosialoside clusters showed that some of the glycodendrimers
Yifan Dong et al.
Carbohydrate polymers, 221, 37-47 (2019-06-23)
Bile salts tend to form micelles in aqueous media and can thereby contribute to drug solubilization; they also exhibit crystallization inhibition properties that can stabilize supersaturated drug solutions. Herein, we explore conjugation of bile salts with polysaccharides to create new
Yonghong Gan et al.
The Journal of organic chemistry, 71(25), 9487-9490 (2006-12-02)
Syntheses of new benzophenone-containing fatty acids (FABPs) 1, 5, and 6 and a new route to FABP 3 are described. Combined with the known 2 and 4, these FABPs comprise a set of photoactivatable fatty acid analogues with the crosslinking
M Adamczyk et al.
Steroids, 62(12), 771-775 (1998-01-22)
Alkylation of 3,17 beta-bis(2-trimethylsilyl)ethoxymethyl-1,3,5(10) estratriene-6-one (2) with 5-bromo-1-pentene using NaHMDS in THF afforded 3,17 beta-bis(2-trimethylsilyl)ethoxymethyl-7-alpha-(4'pentenyl)-1,3,5(10) estratriene-6-one (3) in excellent stereoselectivity (> 95% epimeric excess). Functionalization of the side chain in compound 3 was accomplished via ozonolysis, oxidation and esterification to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service