Skip to Content
Merck
All Photos(1)

Key Documents

774405

Sigma-Aldrich

(S)-2-Aminobutane-1,4-dithiol hydrochloride

99% (titration)

Synonym(s):

(S)-2-Aminobutane-1,4-dithiol hydrochloride, (2S)-2-Amino-1,4-dimercaptobutane hydrochloride, DTBA, Dithiobutylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NS2 · HCl
CAS Number:
Molecular Weight:
173.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99% (titration)

form

solid

reaction suitability

reagent type: reductant

mp

210-225 °C

storage temp.

2-8°C

SMILES string

Cl.N[C@H](CS)CCS

InChI

1S/C4H11NS2.ClH/c5-4(3-7)1-2-6;/h4,6-7H,1-3,5H2;1H/t4-;/m0./s1

InChI key

HWWPXJZINVJNBM-WCCKRBBISA-N

Application

(S)-2-Aminobutane-1,4-dithiol hydrochloride (DTBA), also known as dithiobutylamine, is a biological reducing reagent with enhanced properties over other commonly used reagents. DTBA was shown to reduce small molecule disulfides 3-5x faster than DTT and was able to reduce (activate) the cysteine dependent protease papain 14x faster than DTT.[1][2] In addition, due to the presence of the primary amine group, removal of DTBA from mixtures after use is simplified and can be achieved using cation exchange resin.[1]
Dithiobutylamine or DTBA is a new biological reducing reagent with enhanced properties over other commonly used reagents. DTBA was shown to reduce small molecule disulfides 3-5x faster than DTT and was able to reduce (activate) the cysteine dependent protease papain 14x faster than DTT. In addition, due to the presence of the primary amine group, removal of DTBA from mixtures after use is simplified and can be achieved using cation exchange resin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Troyano et al.
Inorganic chemistry, 58(5), 3290-3301 (2019-02-13)
Direct reactions under ambient conditions between CuX (X = Br, I) and thiobenzamide (TBA) were carried out at different ratios, giving rise to the formation of a series of one-dimensional (1D) coordination polymers, (CPs) [CuI(TBA)] n (1), [Cu3I3(TBA)2] n (4)
A potent, versatile disulfide-reducing agent from aspartic acid.
Lukesh III, et al.
Journal of the American Chemical Society, 134(9), 4057-4059 (2012)
Mozhdeh Imaninezhad et al.
Bioconjugate chemistry, 30(1), 34-46 (2018-12-19)
Macroporous cell-laden hydrogels have recently gained recognition for a wide range of biomedical and bioengineering applications. There are various approaches to create porosity in hydrogels, including lyophilization or foam formation. However, many do not allow a precise control over pore
Coordination properties of dithiobutylamine (DTBA), a newly introduced protein disulfide reducing agent.
Adamczyk J, et al.
Inorganic Chemistry, 54(2), 596-606 (2014)
Aiping Wen et al.
Materials science & engineering. C, Materials for biological applications, 111, 110759-110759 (2020-04-14)
Ovarian cancer is considered to be the most fatal reproductive cancers. Melphalan is used to treat ovarian cancer as an intraperitoneal chemotherapy agent. However, elucidating its pharmacokinetic behavior and preparing it for administration are challenging since it undergoes spontaneous hydrolysis.

Related Content

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Raines lab innovations include traceless Staudinger ligation and DTBA reagent, advancing chemical biology research with Sigma-Aldrich.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service