Skip to Content
Merck
All Photos(1)

Key Documents

713538

Sigma-Aldrich

Synperonic® PE/P84

Synonym(s):

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

form

paste

Quality Level

impurities

≤1.0% water
≤1.0%

solubility

H2O: 0.25 g/10 mL, clear, colorless

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Synperonic is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianan Shen et al.
Biomaterials, 34(5), 1581-1590 (2012-11-14)
The metastasis of breast cancer is the leading cause of cancer death in women, and the lung is a common location of a secondary tumor that has metastasized from the primary source tumor. In this work, an attempt to simultaneously
Tsz Chung Lai et al.
Colloids and surfaces. B, Biointerfaces, 99, 27-37 (2011-10-18)
Polyplex particles formed with plasmid DNA (pDNA) and Pluronic P85-block-poly{N-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (P85-b-P[Asp(DET)]) demonstrated highly effective transfection ability compared to PEG-based block cationomer, PEG-b-P[Asp(DET)]. Ternary polyplexes comprising PEG-b-P[Asp(DET)], poly(ethylene oxide)-b-poly(propylene oxide)-b-poly(ethylene oxide)-b-P[Asp(DET)] (P(EPE)-b-P[Asp(DET)]) used as an analog of P85-b-P[Asp(DET)], and pDNA were
A N Sklifas et al.
Biofizika, 57(2), 317-324 (2012-05-19)
The adsorption abilities of the perfluorocarbon emulsion stabilized by Proxanol 268 were investigated in vitro and in vivo. In vitro, the saturation point for the blood plasma proteins was nearly reached after five minutes of incubation of the emulsion with
Xijun Zhang et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 31(6), 842-845 (2011-12-17)
This study examined the effect of P85 (a pluronic block copolymer) and microbubble (MB) ultrasound contrast agents under ultrasound irradiation on gene transfection and expression. The pEGFP plasmids that can encode enhanced green fluorescent protein (pEGFP) served as a report
Houjian Gong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13590-13600 (2012-09-08)
The block polyethers PEO-PPO-ph-PPO-PEO (BPE) and PPO-PEO-ph-PEO-PPO (BEP) are synthesized by anionic polymerization using bisphenol A as initiator. Compared with Pluronic P123, the aggregation behaviors of BPE and BEP at an air/water interface are investigated by the surface tension and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service