Skip to Content
Merck
All Photos(1)

Key Documents

435406

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)

average Mn ~1,100

Synonym(s):

Pluronic® L-31, PEG-PPG-PEG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor density

>1 (vs air)

vapor pressure

<0.3 mmHg ( 20 °C)

form

viscous liquid

mol wt

average Mn ~1,100

composition

PEG, 10 wt. %

refractive index

n20/D 1.453

surface tension

47 dyn/cm, 25 °C, 0.1 wt. % in H2O

viscosity

175 cP(25 °C, Brookfield)(lit.)

transition temp

softening point −32 °C
cloud point 37 °C (1 wt. % aqueous solution)

density

1.018 g/mL at 25 °C

HLB

1.0 - 7.0

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Surface active material whose properties are dependent on the hydrophilic (EO)/hydrophobic (PO) ratio. High EO content gives better o/w stabilizers.

Physical form

Terminal primary dihydroxy-functional oligomer.

Legal Information

Pluronic is a registered trademark of BASF

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsz Chung Lai et al.
Colloids and surfaces. B, Biointerfaces, 99, 27-37 (2011-10-18)
Polyplex particles formed with plasmid DNA (pDNA) and Pluronic P85-block-poly{N-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (P85-b-P[Asp(DET)]) demonstrated highly effective transfection ability compared to PEG-based block cationomer, PEG-b-P[Asp(DET)]. Ternary polyplexes comprising PEG-b-P[Asp(DET)], poly(ethylene oxide)-b-poly(propylene oxide)-b-poly(ethylene oxide)-b-P[Asp(DET)] (P(EPE)-b-P[Asp(DET)]) used as an analog of P85-b-P[Asp(DET)], and pDNA were
Laurien Buitinga et al.
Clinical and experimental rheumatology, 29(5), 783-789 (2011-10-04)
The Time Trade-Off (TTO) is an instrument used for valuing health-related quality of life. This study evaluated the test-retest reliability of a computer TTO in rheumatoid arthritis (RA), and compared the computer with the interview TTO regarding feasibility and agreement.
Houjian Gong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13590-13600 (2012-09-08)
The block polyethers PEO-PPO-ph-PPO-PEO (BPE) and PPO-PEO-ph-PEO-PPO (BEP) are synthesized by anionic polymerization using bisphenol A as initiator. Compared with Pluronic P123, the aggregation behaviors of BPE and BEP at an air/water interface are investigated by the surface tension and
Jun Song et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 66(4), 792-798 (2012-07-07)
A novel method for the determination of nickel was established by ultrasonic-assisted cloud point extraction (UA-CPE) prior to flame atomic absorption spectrometry (FAAS) analysis. The nickel reacted with N,N'-bis(salicylidene)-1,2-ethanediamine (BSE) to form hydrophobic chelates, which were extracted into the micelles
Wei Xu et al.
Drug delivery, 19(4), 208-219 (2012-05-31)
The aim of this study was to construct novel targeting polymeric micelles. Folate-Poly (ethylenimine)-Pluronic copolymers were synthesized. A paclitaxel (PTX)-loaded mixed micelles consisting of Folate-Poly (ethylenimine)-Pluronic and Pluronic L121 copolymers have been developed. The mixed micelles showed nano-sized spherical morphology.

Articles

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Questions

  1. Can you send me please diffractogram of sample: PEG-block-PPG-Block-PEG 1100

    1 answer
    1. Unfortunately, a diffractogram is not available for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service