Merck
  • Bioreducible polyether-based pDNA ternary polyplexes: balancing particle stability and transfection efficiency.
Bioreducible polyether-based pDNA ternary polyplexes: balancing particle stability and transfection efficiency.
Colloids and surfaces. B, Biointerfaces (2011-10-18)
Tsz Chung Lai, Kazunori Kataoka, Glen S Kwon
ABSTRACT
Polyplex particles formed with plasmid DNA (pDNA) and Pluronic P85-block-poly{N-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (P85-b-P[Asp(DET)]) demonstrated highly effective transfection ability compared to PEG-based block cationomer, PEG-b-P[Asp(DET)]. Ternary polyplexes comprising PEG-b-P[Asp(DET)], poly(ethylene oxide)-b-poly(propylene oxide)-b-poly(ethylene oxide)-b-P[Asp(DET)] (P(EPE)-b-P[Asp(DET)]) used as an analog of P85-b-P[Asp(DET)], and pDNA were prepared in this work aiming at maintaining adequate transfection efficiency while solving the stability issues of the P85-b-P[Asp(DET)] polyplexes. Furthermore, a bioreducible P(EPE)-SS-P[Asp(DET)] possessing a redox potential-sensitive disulfide linkage between the P(EPE) polymer and the cationic block was used as a substitute for P(EPE)-b-P[Asp(DET)] during ternary complex formation to investigate whether the transfection ability of the ternary polyplex system could be enhanced by triggered release of P(EPE) polymers from the polyplexes. The ternary complexes showed significant improvement in terms of stability against salt-induced aggregation compared to binary complexes, although the gene delivery ability dropped with the amount of PEG-b-P[Asp(DET)] used for complexation. By manipulating the difference in redox potential between the extracellular and intracellular environments, the reducible ternary complexes achieved higher transfection compared to the non-reducible polyplexes; moreover, the reducible polyplexes exhibited comparable stability to the non-reducible ones. These results suggest that reducible ternary complexes could provide satisfactory transfection efficiency without comprising the colloidal stability of the particles.
Sigma-Aldrich
Poloxamer 407, micronized
Pricing and availability is not currently available.
Sigma-Aldrich
Poloxamer 188
Pricing and availability is not currently available.
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Pricing and availability is not currently available.
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Poloxamer 188, solid
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Pricing and availability is not currently available.
Sigma-Aldrich
Synperonic® PE P105, surfactant
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Pricing and availability is not currently available.
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Pricing and availability is not currently available.
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Pricing and availability is not currently available.
Sigma-Aldrich
Synperonic® PE/P84
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Poloxamer 188, micronized
Pricing and availability is not currently available.