Skip to Content
Merck
All Photos(1)

Key Documents

576883

Sigma-Aldrich

Tin

nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis

Synonym(s):

Metallic tin, Silver Matt Powder, Tin Powder, Tin element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99% trace metals basis

form

nanopowder

resistivity

11 μΩ-cm, 20°C

particle size

<150 nm (SEM)

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

Application

Tin nanopowder can be used in the fabrication of a variety of devices, such as:
  • lithium ion batteries[1][2]
  • dye sensitized solar cells (DSSCs)[3]
  • fuel cells[4]
  • supercapacitors[5]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-cost copper zinc tin sulfide counter electrodes for high-efficiency dye-sensitized solar cells
Xin X, et al.
Angewandte Chemie (International Edition in English), 50(49), 11739-11742 (2011)
Tin-nanoparticles encapsulated in elastic hollow carbon spheres for high-performance anode material in lithium-Ion batteries
Zhang W, et al.
Advanced Materials, 20(6), 1160-1165 (2008)
A one-step approach towards carbon-encapsulated hollow tin nanoparticles and their application in lithium batteries
Cui G, et al.
Small, 3(12), 2066-2069 (2007)
Titanium nitride nanoparticles based electrocatalysts for proton exchange membrane fuel cells
Avasarala B, et al.
Journal of Materials Chemistry, 19(13), 1803-1805 (2009)
Stabilized TiN nanowire arrays for high-performance and flexible supercapacitors
Lu X, et al.
Nano Letters, 12(10), 5376-5381 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service