Skip to Content
Merck
All Photos(4)

Key Documents

484059

Sigma-Aldrich

Silver

nanopowder, <150 nm particle size, 99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99% trace metals basis

form

nanopowder

resistivity

1.59 μΩ-cm, 20°C

particle size

<150 nm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

Silver nanoparticles (Ag-NPs) have unique physical and chemical properties such as high thermal and electrical conductivity, chemical stability, catalytic activity, surface-enhanced Raman scattering, and nonlinear optical behavior. Ag-NPs are widely used in the fields of electronics, catalysis, and medicine.[1]

Application

Silver can be used as a catalyst for the direct electroreduction ofcarbon dioxide.[2]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eun-Jung Park et al.
Archives of pharmacal research, 34(2), 299-307 (2011-03-08)
The adverse effects of silver nanoparticles (AgNPs) to human and environment have not been known well, although AgNPs are now widely applied to consumer products. In this study, we investigated the inflammatory responses including cytokine production and gene expression in
Eun-Jung Park et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(3), 872-878 (2009-12-09)
Silver nanoparticles (AgNPs) are widely applied in many household products and medical uses. However, studies on the effects of AgNPs on human health and environmental implications are in the beginning stage. Furthermore, most data on the toxicity of AgNPs have
Highly selective and scalable CO2 to CO - Electrolysis using coral-nanostructured Ag catalysts in zero-gap configuration
Woong Hee Lee, et al.
Nano Energy, 76, 105030-105030 (2020)
Eun-Jung Park et al.
Environmental toxicology and pharmacology, 30(2), 162-168 (2011-07-27)
Toxicity of nanoparticles depends on many factors including size, shape, chemical composition, surface area, surface charge, and others. In this study, we compared the toxicity of different sized-silver nanoparticles (AgNPs) which are being widely used in consumer products due to
Effect of nanoparticle stabilization and physicochemical properties on exposure outcome: acute toxicity of silver nanoparticle preparations in zebrafish (Danio rerio)
Cunningham S, et al.
Environmental Science & Technology, 47(8), 3882-3892 (2013)

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service