Skip to Content
Merck
All Photos(2)

Key Documents

537349

Sigma-Aldrich

Ethyl acetoacetate

ReagentPlus®, 99%

Synonym(s):

EAA, Ethyl 3-oxobutanoate, Acetoacetic ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC2H5
CAS Number:
Molecular Weight:
130.14
Beilstein:
385838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.48 (vs air)

vapor pressure

1 mmHg ( 28.5 °C)

product line

ReagentPlus®

Assay

99%

autoignition temp.

580 °F

expl. lim.

9.5 %

bp

181 °C (lit.)

mp

−43 °C (lit.)

solubility

water: soluble 130 g/L at 20 °C

density

1.029 g/mL at 20 °C (lit.)

functional group

ester
ketone

SMILES string

CCOC(=O)CC(C)=O

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

XYIBRDXRRQCHLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ethyl acetoacetate is a versatile reagent that can be used as a nucleophile in alkylation, conjugate addition and condensation reactions. Some of its applications are:
  • Alkylation at the α-carbon of ethyl acetoacetate followed by hydrolysis and decarboxylation can afford a variety of methyl ketones.
  • It also undergoes acylation at the α-carbon in the presence of MgCl2 and pyridine to give synthetically important intermediates.
  • It can be used in Knoevenagel condensation with aliphatic, aromatic, and heteroaromatic aldehydes to produce α-alkylideneacetoacetates.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

164.3 °F - closed cup

Flash Point(C)

73.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knoevenagel-kondensationen mit titantetrachlorid/base?II: Alkyliden-und arylidenacet-bzw.-nitroessigester bei 0?22?.
Lehnert W
Tetrahedron, 28(3), 663-666 (1972)
Synthetic Applications of Dealkoxycarbonylations of Malonate Esters, β-Keto Esters, α-Cyano Esters and Related Compounds in Dipolar Aprotic Media-Part II.
Krapcho A P
Synthesis, 1982(11), 893-914 (1982)
Ethyl Acetoacetate.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Procedures for the acylation of diethyl malonate and ethyl acetoacetate with acid chlorides using tertiary amine bases and magnesium chloride.
Rathke MW and Cowan PJ.
The Journal of Organic Chemistry, 50(15), 2622-2624 (1985)
Gui-Rong Qu et al.
Organic letters, 11(8), 1745-1748 (2009-03-20)
A novel approach to the synthesis of purines bearing functionalized carbon substituents or methyl in position 6 was developed. Under different reaction conditions, 6-halopurine derivatives could react with ethyl acetoacetate efficiently to yield 2-(purin-6-yl)acetoacetic acid ethyl esters, (purin-6-yl)acetates and 6-methylpurines

Articles

The Biginelli Reaction is an acid-catalyzed, three-component reaction between an aldehyde, b-ketoester, and urea that produces tetrahydropyrimidones, which have potential pharmaceutical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service