Skip to Content
Merck
All Photos(2)

Key Documents

524417

Sigma-Aldrich

3,5-Dinitro-o-toluamide

98%

Synonym(s):

2-Methyl-3,5-dinitrobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H2(CH3)CONH2
CAS Number:
Molecular Weight:
225.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

170-180 °C (lit.)

functional group

amide
nitro

SMILES string

Cc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C8H7N3O5/c1-4-6(8(9)12)2-5(10(13)14)3-7(4)11(15)16/h2-3H,1H3,(H2,9,12)

InChI key

ZEFNOZRLAWVAQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Water solubility of 3,5-Dinitro-o-toluamide (3,5-DNoTAME) is 447.5mg/L. It affords 3-amino-5-nitro-o-toluamide, via biological reduction. 3,5-DNoTAME also forms colored complexes on reaction with various diamines and organic solvents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anticoccidial drugs: effects on infectivity and survival intracellularly of Eimeria tenella sporozoites.
L R McDougald et al.
Experimental parasitology, 40(3), 314-319 (1976-12-01)
S J Ball et al.
Parasitology research, 73(4), 293-297 (1987-01-01)
Medication of chicks with 125 ppm amprolium or dinitolmide adversely affected oocyst sporulation of Eimeria acervulina (Weybridge strain). Dinitolmide delayed oocyst production and no oocyst wall formation was seen up to 168 h post infection. Both drugs caused large numbers
T Ohta et al.
Mutation research, 77(1), 21-30 (1980-01-01)
18 feed additives were tested for DNA-modifying effects by the repair test named "rec-assay" with Bacillus subtillis H17 (rec+) and M45 (rec-), and for mutagenicity with Escherichia coli WP2 hcr and 5 Salmonella typhimurium tester strains with the use of
L P Joyner et al.
Parasitology, 75(2), 155-164 (1977-10-01)
Chicks infected with the Weybridge strains of Eimeria maxima and E. acervulina were not protected by the normal levels of amprolium, but the sporulation of the oocysts was inhibited. With higher drug concentrations, fewer oocysts were produced. E brunetti was
Feed Additive Residues, Isolation and Identification of Amino-nitro-o-toluamide Formed by Biological Reduction of 3, 5-Dinitro-o-tuamide.
Smith GN, et al.
Journal of Agricultural and Food Chemistry, 11(3), 257-260 (1963)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service