Skip to Content
Merck
All Photos(4)

Key Documents

517453

Sigma-Aldrich

Copper(II) acetate

greener alternative

powder, 99.99% trace metals basis

Synonym(s):

Cupric acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(CO2CH3)2
CAS Number:
Molecular Weight:
181.63
Beilstein:
3595638
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.9 (vs air)

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

core: copper

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

CC(=O)O[Cu]OC(C)=O

InChI

1S/2C2H4O2.Cu/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

OPQARKPSCNTWTJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper acetate (Cu (OAc)2) occurs in dimeric structure in solid state and in non-aqueous solution. Anhydrous Cu (OAc)2 may be manufactured by heating or refluxing the monohydrate Cu (OAc)2 at 90oC and then washing the insoluble product with diethyl ether.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Copper acetate forms complexes which may be used in biochemical reactions such as; DNA extraction, binding ribose hydroxyls of purine and pyrimidine nucleosides. A study has shown Cu (OAc)2 to fix and remove tannins in samples from Pyrus yielding high quality DNA. The complexes have been studied for anticancer, anti-bacterial and antifungal activities.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manpreet Kaur et al.
Nanoscale research letters, 13(1), 182-182 (2018-06-20)
Here, we report efficient composition-tunable Cu-doped ZnInS/ZnS (core and core/shell) colloidal nanocrystals (CNCs) synthesized by using a colloidal non-injection method. The initial precursors for the synthesis were used in oleate form rather than in powder form, resulting in a nearly
Michael Devereux et al.
Journal of inorganic biochemistry, 98(6), 1023-1031 (2004-05-20)
Thiabendazole (TBZH) reacts with iron(III) nitrate causing protonation of the ligand to yield the nitrate salt [TBZH(2)NO(3)] (1). Reaction of TBZH with copper(II) acetate results in the deprotonation of the ligand yielding [Cu(TBZ)2.(H2O)2] (2). Reactions of TBZH with the chloride
Stereoselective differentiation between ribonucleosides and deoxynucleosides by reaction with the copper(II) acetate dimer.
N A Berger et al.
Nature: New biology, 239(95), 237-240 (1972-10-25)
Quantitative densitometric thin-layer chromatography of lipids using copper acetate reagent.
M E Fewster et al.
Journal of chromatography, 43(1), 120-126 (1969-08-05)
Sarayut Radapong et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-05)
Artocarpus lakoocha Wall. ex Roxb. (family: Moraceae) has been used as a traditional Thai medicine for the treatment of various parasitic diseases. This species has been reported to be the source of phytochemicals, which show potent biological activities. The objective

Articles

Copper metal deposition processes are an essential tool for depositing interconnects used in microelectronic applications, giving group 11 (coinage metals: Copper, Silver, and Gold) an important place in atomic layer deposition (ALD) process development.

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service