Skip to Content
Merck
All Photos(2)

Key Documents

403342

Sigma-Aldrich

Copper(I) acetate

97%

Synonym(s):

Copper monoacetate, Cuprous acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCO2CH3
CAS Number:
Molecular Weight:
122.59
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

powder and chunks

reaction suitability

core: copper
reagent type: catalyst

mp

250 °C (dec.) (lit.)

SMILES string

CC(=O)O[Cu]

InChI

1S/C2H4O2.Cu/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

RFKZUAOAYVHBOY-UHFFFAOYSA-M

Application

Copper acetate(CuOAc) was used as a starting material for the preparation of copper nanoparticles and CuAlO2 p-type nanostructured semiconductors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of delafossite CuAlO 2 p-type semiconductor with a nanoparticle-based Cu (I) acetate-loaded boehmite precursor
Thu TV, et al.
Mat. Res. Bul., 46.11, 1819-1827 (2011)
Room?Temperature Formation of Hollow Cu2O Nanoparticles.
Hung LI, et al.
Advanced Materials, 22.17, 1910-1914 (2010)
Thomas G Meikle et al.
ACS applied materials & interfaces, 12(6), 6944-6954 (2020-01-10)
Herein, we demonstrate a method for the functionalization of cubic phase lipid nanoparticles (cubosomes) with a series of magnetite (Fe3O4), copper oxide (Cu2O), and silver (Ag) nanocrystals, with prospective applications across a wide range of fields, including antimicrobial treatments. The
Jan Lorkowski et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(48), 11365-11374 (2019-07-06)
Cyclic (amino)(aryl)carbenes (cAArCs) based on the isoindoline core were successfully generated in situ by α-elimination of 3-alkoxyisoindolines at high temperatures or by deprotonation of isoindol-2-ium chlorides with sodium or copper(I) acetates at low temperatures. 3-Alkoxy-isoindolines 2 a,b-OR (R=Me, Et, iPr) have
You Zhai et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 17(5), 741-751 (2015-11-04)
Cu2 S/ZnS heterostructured nanorods (HNRs) with uncommon morphologies are achieved through single-pot and multi-batch synthetic strategies. In both cases, Cu2 S NRs form first, which then undergo partial cation exchange and solution-liquid-solid (SLS)-like growth catalyzed by the remaining Cu2 S

Articles

Copper metal deposition processes are an essential tool for depositing interconnects used in microelectronic applications, giving group 11 (coinage metals: Copper, Silver, and Gold) an important place in atomic layer deposition (ALD) process development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service