Skip to Content
Merck
All Photos(2)

Key Documents

493678

Sigma-Aldrich

Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate

98%

Synonym(s):

Decamethylcobaltocenium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30CoF6P
CAS Number:
Molecular Weight:
474.35
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

reaction suitability

core: cobalt
reagent type: catalyst

mp

305 °C (dec.) (lit.)

SMILES string

[Co+].F[P-](F)(F)(F)(F)F.C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/2C10H15.Co.F6P/c2*1-6-7(2)9(4)10(5)8(6)3;;1-7(2,3,4,5)6/h2*1-5H3;;/q;;+1;-1

InChI key

ILKSSYYMFRGBHO-UHFFFAOYSA-N

Application

Reactant for:
  • Spontaneous assembly of titanium and gallium bis(dihydroxybenzamido)naphthalene supramolecular tetranuclear cluster complexes
  • Preparation of cobaltocenium nickel bisdithiolene complexes
  • Metathesis reaction with potassium biphenoxide
  • Nucleophilic addition and single-electron reduction reactions

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service