This product is not tested for solubility. However, various sources note that this compound is soluble in dry acetone, dimethylformamide, and hot ethanol. No rate has been reported.
279811
Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate
98%
Synonym(s):
Cobaltocenium hexafluorophosphate
About This Item
Recommended Products
Quality Level
Assay
98%
reaction suitability
core: cobalt
reagent type: catalyst
SMILES string
[Co+].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.F[P-](F)(F)(F)(F)F
InChI
1S/2C5H5.Co.F6P/c2*1-2-4-5-3-1;;1-7(2,3,4,5)6/h2*1-5H;;/q;;+1;-1
InChI key
IASMSXQWVNBFOM-UHFFFAOYSA-N
Application
- Cocatalyst for Heck reaction of aryl iodides with alkenes using palladium as catalyst in water
- Reactant for modification of organometallic electrodes surfaces through cathodic reduction
- Reactant in synthesis and solution self-assembly of side-chain cobaltocenium-containing block copolymers
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the solubility?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service