Skip to Content
Merck
All Photos(2)

Key Documents

491047

Sigma-Aldrich

2-(Bromomethyl)pyridine hydrobromide

98%

Synonym(s):

(2-Pyridyl)methyl bromide hydrobromide, 2-Bromomethylpyridine monohydrobromide, 2-Picolyl bromide-hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrN · HBr
CAS Number:
Molecular Weight:
252.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

149-152 °C (lit.)

functional group

bromo

SMILES string

Br[H].BrCc1ccccn1

InChI

1S/C6H6BrN.BrH/c7-5-6-3-1-2-4-8-6;/h1-4H,5H2;1H

InChI key

JQDNCGRNPYKRAO-UHFFFAOYSA-N

General description

2-(Bromomethyl)pyridine hydrobromide is a pyridine derivative. It participates in the synthesis of 7-nitrobenz-2-oxa-1,3-diazole (NBD) based colorimetric and fluorescence chemosensor.

Application

2-(Bromomethyl)pyridine hydrobromide may be used in the preparation of :
  • 2-[1-(pyridin-2-ylmethyl)-1H-1,2,3-triazol-4-yl]pyridine
  • 2-morpholin-4-yl-7-(pyridin-2-ylmethoxy)-4H-1,3-benzoxazin-4-one
  • 8-methyl-2-morpholin-4-yl-7-(pyridin-2-ylmethoxy)-4H-1,3-benzoxazin-4-one
  • trans-4-(4-(bis(pyridin-2-ylmethyl)amino)styryl)benzonitrile
  • 2-[N-(2-aminoethanethiol) methyl]-pyridine
  • 2-[N-(ethylenediamino)-methyl]-pyridine
  • 3-(carboxymethyl)-1-[(pyridin-2-yl)methyl]-3H-imidazol-1-ium trifluoroacetate
  • 3-[(S)-1-carboxy-2-methylpropyl]-1-[(pyridin-2-yl)methyl]-3H-imidazol-1-ium trifluoroacetate

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An NBD-based colorimetric and fluorescent chemosensor for Zn2+ and its use for detection of intracellular zinc ions.
Xu Z, et al.
Tetrahedron, 65(11), 2307-2312 (2009)
Synthesis and characterization of platinum (II) complexes from trifluoromethyl phenylenediamine, picoline and N-benzyl ethylenediamine derivatives.
Almeida MVD, et al.
Journal of the Brazilian Chemical Society, 17(7), 1266-1273 (2006)
Green Catalysts: Solid-Phase Peptide Carbene Ligands in Aqueous Transition-Metal Catalysis.
Worm-Leonhard K and Meldal M.
European Journal of Organic Chemistry, 2008(31), 5244-5253 (2008)
A stilbene-based fluoroionophore for copper ion sensing in both reduced and oxidized environments.
Zhu MQ, et al.
Talanta, 81(1), 678-683 (2010)
A multicomponent CuAAC ?click? approach to a library of hybrid polydentate 2-pyridyl-1,2,3-triazole ligands: new building blocks for the generation of metallosupramolecular architectures.
Crowley JD and Bandeen PH.
Dalton Transactions, 39(2), 612- 623 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service