162701
2-(Chloromethyl)pyridine hydrochloride
98%
Synonym(s):
2-Picolyl chloride hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H6ClN · HCl
CAS Number:
Molecular Weight:
164.03
Beilstein:
3689155
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
120-124 °C (lit.)
functional group
chloro
SMILES string
Cl[H].ClCc1ccccn1
InChI
1S/C6H6ClN.ClH/c7-5-6-3-1-2-4-8-6;/h1-4H,5H2;1H
InChI key
JPMRGPPMXHGKRO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Selective sensing of zinc ions with a novel magnetic resonance imaging contrast agent.
Hanaoka K, et al.
J. Chem. Soc. Perkin Trans. II, 9, 1840-1843 (2001)
Functionalization of p-tert-Butylcalix [5] arene by Alkylation with 2-(Chloromethyl) pyridine Hydrochloride.
Pappalardo S and Ferguson G.
The Journal of Organic Chemistry, 61(7), 2407-2412 (1996)
Functionalization of p-tert-butylcalix [6] arene by alkylation with 2-(chloromethyl) pyridine hydrochloride.
Neri P and Pappalardo S.
The Journal of Organic Chemistry, 58(5), 1048-1053 (1993)
M Batiste-Alentorn et al.
Mutation research, 341(3), 161-167 (1995-01-01)
To provide further background data for the somatic mutation and/or recombination tests in Drosophila melanogaster, we have evaluated the response in 3 assays (zeste-white, white-ivory and wing spot) of 5 chemicals classified by the U.S. National Toxicology Program (NTP) as
Alexander R Parent et al.
Dalton transactions (Cambridge, England : 2003), 43(33), 12501-12513 (2014-07-09)
Iron tris(2-methylpyridyl)amine (TPA) and iron 1-(bis(2-methylpyridyl)amino)-2-methyl-2-propanoate (BPyA) salts are characterized as water oxidation catalysts (WOCs) using sodium periodate. Under the conditions used, these complexes serve as homogeneous WOCs as demonstrated via kinetic analysis and dynamic light scattering (DLS). The Fe(BPyA)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service