Skip to Content
Merck
All Photos(1)

Key Documents

482609

Sigma-Aldrich

N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine

97%

Synonym(s):

([2S,3R]-3-Amino-2-hydroxy-4-phenylbutanoyl)-L-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH[NHCOCH(OH)CH(NH2)CH2C6H5]CO2H
CAS Number:
Molecular Weight:
308.37
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

powder or crystals

optical activity

[α]20/D −11°, c = 1 in 1 M NaOH

reaction suitability

reaction type: solution phase peptide synthesis

mp

245 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)C[C@H](NC(=O)[C@@H](O)[C@H](N)Cc1ccccc1)C(O)=O

InChI

1S/C16H24N2O4/c1-10(2)8-13(16(21)22)18-15(20)14(19)12(17)9-11-6-4-3-5-7-11/h3-7,10,12-14,19H,8-9,17H2,1-2H3,(H,18,20)(H,21,22)/t12-,13+,14+/m1/s1

InChI key

VGGGPCQERPFHOB-RDBSUJKOSA-N

Gene Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela P Alves et al.
Pharmacology, 89(1-2), 22-28 (2012-01-13)
The aim of the present study was to investigate the mechanisms underlying the endogenous control of nociception at a peripheral level during inflammation. Using a pharmacological approach and the rat paw pressure test, we assessed the effect of an intraplantar
Xiaopan Zhang et al.
Journal of enzyme inhibition and medicinal chemistry, 28(3), 545-551 (2012-03-03)
Aminopeptidase N (APN/CD13) over expressed on tumour cells, plays a critical role in tumour invasion, metastasis and tumour angiogenesis. In this article, we described the design, synthesis and preliminary activity studies of novel 3-amino-2-hydroxyl-3-phenylpropanoic acid derivatives as APN inhibitors. The
Kunio Fujiwara et al.
Journal of molecular histology, 42(6), 589-596 (2011-10-19)
The in vivo role of transporters in drug disposition, in the context of other transporters, and metabolism has not been established. We prepared an anti-bestatin serum against bestatin conjugated to albumin with glutaraldehyde (GA). The antiserum was specific for GA-conjugated
Yekaterina Poloz et al.
Eukaryotic cell, 11(4), 545-557 (2012-02-22)
Bestatin methyl ester (BME) is an inhibitor of Zn(2+)-binding aminopeptidases that inhibits cell proliferation and induces apoptosis in normal and cancer cells. We have used Dictyostelium as a model organism to study the effects of BME. Only two Zn(2+)-binding aminopeptidases
Atsushi Wakita et al.
International journal of hematology, 96(1), 84-93 (2012-05-29)
We conducted a multicenter prospective randomized study to compare a fixed-scheduled induction therapy with a response-oriented individualized induction therapy for elderly patients with acute myeloid leukemia (AML). Newly diagnosed AML patients, aged between 65 and 80, were randomly assigned to

Questions

  1. What is the recommended method for storing stock solutions of 4826 the product 09, N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine?

    1 answer
    1. Stock solutions are stable for up to 1 month at -20°C

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service