Merck
  • Calcium plus vitamin D supplementation and health outcomes five years after active intervention ended: the Women's Health Initiative.
Calcium plus vitamin D supplementation and health outcomes five years after active intervention ended: the Women's Health Initiative.
Journal of women's health (2002) (2013-10-18)
Jane A Cauley, Rowan T Chlebowski, Jean Wactawski-Wende, John A Robbins, Rebecca J Rodabough, Zhao Chen, Karen C Johnson, Mary Jo O'Sullivan, Rebecca D Jackson, JoAnn E Manson
ABSTRACT
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were randomized; postintervention follow-up continued among 29,862 (86%) of surviving participants. Primary outcomes were hip fracture and colorectal cancer. Breast cancer, all cancers, cardiovascular disease (CVD), and total mortality were predetermined major study outcomes. Hip fracture incidence was comparable in the supplement and the placebo groups, postintervention hazard ratio (HR)=0.95, 95% confidence interval (95% CI: 0.78, 1.15) and overall HR=0.91 (95% CI: 0.79, 1.05). Overall, colorectal cancer incidence did not differ between randomization groups, HR=0.95 (95% CI: 0.80, 1.13). Throughout, there also was no difference in invasive breast cancer, CVD, and all-cause mortality between groups. In subgroup analyses, the invasive breast cancer effect varied by baseline vitamin D intake (p=0.03 for interaction). Women with vitamin D intakes >600 IU/d, had an increased risk of invasive breast cancer, HR=1.28 (95% CI; 1.03, 1.60). Over the entire study period, in post hoc analyses, the incidence of vertebral fractures, HR=0.87 (95% CI: 0.76, 0.98) and in situ breast cancers, HR=0.82 (95% CI: 0.68, 0.99) were lower among women randomized to supplementation. After an average of 11 years, calcium and vitamin D supplementation did not decrease hip fracture or colorectal cancer incidence. Exploratory analyses found lower vertebral fracture and in situ breast cancer incidence in the supplement users. There was no effect on CVD or all-cause mortality.
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis