Skip to Content
Merck
All Photos(3)

Key Documents

471801

Sigma-Aldrich

4-Pentenoic anhydride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCH2CH2CO)2O
CAS Number:
Molecular Weight:
182.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.447 (lit.)

bp

78-81 °C/0.4 mmHg (lit.)

density

0.997 g/mL at 25 °C (lit.)

SMILES string

C=CCCC(=O)OC(=O)CCC=C

InChI

1S/C10H14O3/c1-3-5-7-9(11)13-10(12)8-6-4-2/h3-4H,1-2,5-8H2

InChI key

NEDHQDYBHYNBIF-UHFFFAOYSA-N

General description

4-Pentenoic anhydride is a carboxylic anhydride.

Application

4-Pentenoic anhydride may be used:
  • In the preparation of glucose functionalized (co)polymers.
  • As a monomer in the preparation of cross-linked polyanhydrides.
  • For the preparation of polymers with pendant vinyl or acetylene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katie L Poetz et al.
Biomacromolecules, 15(7), 2573-2582 (2014-05-23)
Several critical aspects of cross-linked polyanhydrides made using thiol-ene polymerization are reported, in particular the erosion, release, and solution properties, along with their cytotoxicity toward fibroblast cells. The monomers used to synthesize these polyanhydrides were 4-pentenoic anhydride and pentaerythritol tetrakis(3-mercaptopropionate).
Marlène Rippe et al.
Biomaterials science, 7(7), 2850-2860 (2019-05-10)
Glycosaminoglycans (GAGs) are important components of the extracellular matrix that have attracted great interest for drug delivery and pharmaceutical applications due to their diverse biological functions. Among GAGs, heparosan (Hep), a biosynthetic precursor of heparin, has recently emerged as a
Vien T Huynh et al.
Biomacromolecules, 12(5), 1738-1751 (2011-04-12)
Statistical and block copolymers based on poly(2-hydroxyethyl methacrylate) (PHEMA) and poly[oligo(ethylene glycol) methylether methacrylate] (POEGMEMA) were modified with 4-pentenoic anhydride or 4-oxo-4-(prop-2-ynyloxy)butanoic anhydride to generate polymers with pendant vinyl or acetylene, respectively. Subsequent thiol-ene or thiol-yne reaction with thioglycolic acid
Mime Kobayashi et al.
Journal of biomaterials science. Polymer edition, 30(13), 1161-1171 (2019-06-16)
We have developed biocompatible scaffolds that enable cell fate control with visible light. The scaffolds are based on synthetic collagen-like polypeptide, poly(prolyl-hydroxyprolyl-glycyl) {poly(Pro-Hyp-Gly)} which has been used for cosmetics and other healthcare applications. Bioactive peptides were conjugated to the scaffolds
Studies of the mechanism of the hypoglycemic action of 4-pentenoic acid.
Corredor C, et al.
Proceedings of the National Academy of Sciences of the USA, 58(6), 2299-2299 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service