Skip to Content
Merck
All Photos(1)

Key Documents

114235

Sigma-Aldrich

1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione

98%

Synonym(s):

1,3,5-Tri-2-propen-1-yl-1,3,5-triazine-2,4,6(1H ,3H ,5H )-trione, 1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione, 1,3,5-Triallylisocyanurate, 1,3,5-Triallylisocyanuric acid, 1,3,5-Tris-2′-propenylisocyanuric acid, Triallyl isocyanurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
Beilstein:
225482
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

contains

500 ppm tert-butylhydroquinone as inhibitor

refractive index

n20/D 1.513 (lit.)

bp

149-152 °C/4 mmHg (lit.)

density

1.159 g/mL at 25 °C (lit.)

SMILES string

C=CCN1C(=O)N(CC=C)C(=O)N(CC=C)C1=O

InChI

1S/C12H15N3O3/c1-4-7-13-10(16)14(8-5-2)12(18)15(9-6-3)11(13)17/h4-6H,1-3,7-9H2

InChI key

KOMNUTZXSVSERR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, also known as triallyl isocyanurate (TAIC), is a versatile monomer widely employed in polymer synthesis as a crosslinking agent or co-monomer. It is commonly used as a key component in various applications, including rubber and plastic industries, coatings, adhesives, elastomers, and in the development of dental restorative materials.

Application

1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione can be used as:
  • A crosslinking agent or additive in the synthesis of polylactic acid /flax composite materials to enhance their properties and performance under gamma irradiation.
  • A monomer in the synthesis of flexible ionogels with good mechanical properties via in situ thiol-ene photopolymerization with trimethylolpropane tris(3-mercaptopropionate). These ionogels further find applications in electrochemical capacitors.
  • A monomer in the fabrication of the shape memory polymer substrates.
The Progress in Development of Dental Restorative Materials

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.1 °F - closed cup

Flash Point(C)

159.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingbing Zeng et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 789(2), 265-272 (2003-05-14)
Biocompatible affinity adsorbents prepared from macroporous glycidyl methacrylate-triallyl isocyanurate copolymer (GT) has been used for removing circulating immune complexes (CICs). In this work, adsorption of circulating immune complexes on GT-based affinity adsorbents has been studied by using batch and hemoperfusion
Chuanglong He et al.
Journal of biomedical materials research. Part A, 99(4), 655-665 (2011-09-29)
The radiation crosslinked poly(L-lactide) (PLLA) electrospun nanofibers have been developed with improved thermal stability and mechanical properties. Trially isocyanurate (TAIC) were added into PLLA solution at different weight ratios (1, 3, and 5%) and electrospun into nanofibrous mats, the mats
Bolesław Szadkowski et al.
Materials (Basel, Switzerland), 13(3) (2020-02-06)
Perlite and vermiculite are naturally occurring minerals, commonly used by industry to obtain highly thermoisolative and/or non-flammable materials. However, there has been little research into the preparation and application of rubber compounds containing these inexpensive mineral fillers. Here, we show
Max J Männel et al.
Micromachines, 11(3) (2020-03-01)
Three-dimensional (3D) printing of microfluidic devices continuously replaces conventional fabrication methods. A versatile tool for achieving microscopic feature sizes and short process times is micro-stereolithography (µSL). However, common resins for µSL lack biocompatibility and are cytotoxic. This work focuses on
Christopher L Frewin et al.
Polymers, 11(5) (2019-05-22)
Thiol-ene/acrylate-based shape memory polymers (SMPs) with tunable mechanical and thermomechanical properties are promising substrate materials for flexible electronics applications. These UV-curable polymer compositions can easily be polymerized onto pre-fabricated electronic components and can be molded into desired geometries to provide

Articles

With dentists placing nearly 100 million dental fillings into patients′ teeth annually in the U.S. alone, polymeric composite restoratives account for a very large share of the biomaterials market.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service