Skip to Content
Merck
All Photos(3)

Key Documents

464309

Sigma-Aldrich

Methoxyperfluorobutane

99%, mixture of n- and iso-butyl isomers

Synonym(s):

Nonafluorobutyl methyl ether, HFE-7100, Methoxynonafluorobutane, Methoxyperfluorobutane, Nonafluoromethoxybutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCF2R, R = -CF2CF2CF3 or -CF(CF3)2
CAS Number:
Molecular Weight:
250.06
Beilstein:
8768491
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.3 (lit.)

bp

60 °C (lit.)

mp

−135 °C (lit.)

density

1.52 g/mL at 25 °C (lit.)

SMILES string

COC(F)(F)C(F)(F)C(F)(F)C(F)(F)F.COC(F)(F)C(F)(C(F)(F)F)C(F)(F)F

InChI

1S/2C5H3F9O/c1-15-5(13,14)3(8,9)2(6,7)4(10,11)12;1-15-5(13,14)2(6,3(7,8)9)4(10,11)12/h2*1H3

InChI key

OKIYQFLILPKULA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methoxyperfluorobutane can be used as a solvent:
  • In the synthesis of various aryl fluoride derivatives via electrophilic fluorination of aryl and heteroaryl Grignard reagents.
  • In grafting of fluorinated diblock copolymers onto silica particles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Hsin Han et al.
Langmuir : the ACS journal of surfaces and colloids, 26(9), 6108-6110 (2010-03-31)
Three-dimensional organic microfabrication, an emerging technology, faces the challenge of lacking a sacrificial agent (SA) to temporarily support the formation of microscale geometries, which can be removed after a microstructure is constructed. In this study, an ultradense oil-in-organofluorine colloidal emulsion
Coating of silica particles by fluorinated diblock copolymers and use of the resultant silica for superamphiphobic surfaces
Grozea CM, et al.
Polymer, 64(12), 153-162 (2015)
Alfonso Bentolila et al.
Forensic science international, 278, 374-378 (2017-08-15)
Working in crime scenes presents a challenge to the forensic scientist, as some surfaces, such as floors and walls, cannot be transferred to the lab for further development and must, therefore, be processed at the crime scene itself. Two main
Howard C Knachel et al.
Magnetic resonance in chemistry : MRC, 51(7), 407-413 (2013-05-25)
The 3M Company product Novec™ 71IPA DL, a mixture of methoxyperfluorobutane, methoxyperfluoroisobutane and 4.5 wt.% isopropyl alcohol, has been found to be very stable at ambient temperature, producing fluoride at the rate of ~1 ppm/year. Our earlier kinetic and theoretical studies have
Niko Nicolasora et al.
Forensic science international, 288, 266-277 (2018-05-25)
This paper contains details of work carried out to identify the most effective processing conditions for the optimized 1,2-indandione/zn formulation developed for use under UK conditions. Using direct measurements of fluorescence taken from test spots of amino acids and eccrine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service