Skip to Content
Merck
All Photos(1)

Key Documents

P9900

Sigma-Aldrich

Perfluorodecalin

95%

Synonym(s):

Octadecafluorodecahydronaphthalene (cis+trans), Perflunafene, Perfluorodecahydronaphthalene (cis+trans)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10F18
CAS Number:
Molecular Weight:
462.08
Beilstein:
2067113
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

17.5 (vs air)

Quality Level

Assay

95%

refractive index

n20/D 1.3145 (lit.)

bp

142 °C (lit.)

mp

−10 °C (lit.)

density

1.908 g/mL at 25 °C (lit.)

SMILES string

FC1(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C1(F)F

InChI

1S/C10F18/c11-1-2(12,5(17,18)9(25,26)7(21,22)3(1,13)14)6(19,20)10(27,28)8(23,24)4(1,15)16

InChI key

UWEYRJFJVCLAGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Perfluorodecalin is a fluorous solvent generally used as a primary component of the fluorous biphasic system (FBS) or the fluorous multiphasic system (FMS) in synthetic chemistry. It is also used as an additive to increase oxygen solubility in fermentation media.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F

Flash Point(C)

59 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dilution-free analysis from picoliter droplets by nano-electrospray ionization mass spectrometry.
Ryan T Kelly et al.
Angewandte Chemie (International ed. in English), 48(37), 6832-6835 (2009-08-19)
Three-component one-pot synthesis of pyrimidinone derivatives in fluorous media: Ytterbium bis (perfluorooctanesulfonyl) imide complex catalyzed Biginelli-type reaction.
Hong M and Cai C
Journal of Heterocyclic Chemistry, 46(6), 1430-1432 (2009)
Helen G Pennington et al.
PLoS pathogens, 15(3), e1007620-e1007620 (2019-03-12)
The biotrophic fungal pathogen Blumeria graminis causes the powdery mildew disease of cereals and grasses. We present the first crystal structure of a B. graminis effector of pathogenicity (CSEP0064/BEC1054), demonstrating it has a ribonuclease (RNase)-like fold. This effector is part
Palladium-Catalyzed Oxidation and Cyclization of Carbon-Carbon Triple Bonds in Fluorous Media Using Molecular Oxygen.
Wen Y, et al.
Synlett, 2011(07), 1023-1027 (2011)
Convenient electrophilic fluorination of functionalized aryl and heteroaryl magnesium reagents.
Yamada S, et al.
Angewandte Chemie (International Edition in English), 122(12), 2261-2264 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service