Skip to Content
Merck
All Photos(2)

Key Documents

464236

Sigma-Aldrich

Zirconium(IV) hydrogenphosphate

Synonym(s):

α-Zirconium phosphate, γ-Zirconium phosphate, Zirconium bis(hydrogen phosphate), Zirconium bis(monohydrogen orthophosphate), Zirconium hydrogen orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(HPO4)2
CAS Number:
Molecular Weight:
283.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

Quality Level

reaction suitability

reagent type: catalyst
core: zirconium

density

3.3 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Zr+4].OP([O-])([O-])=O.OP([O-])([O-])=O

InChI

1S/2H3O4P.Zr/c2*1-5(2,3)4;/h2*(H3,1,2,3,4);/q;;+4/p-4

InChI key

QOKYJGZIKILTCY-UHFFFAOYSA-J

Application

Employed in the preparation of layered organic-inorganic, hybrid, conducting materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carissa M Vetromile et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(2), 648-652 (2011-01-11)
In this report the photo-physical properties of 9-amino acridine (9AA) associated with αZr-phosphate particles (αZrP) is examined. In ethanol solution 9AA exhibits absorption maxima at 425 nm, 402 nm and 383 nm as well as emission bands centered at 455
Roxane M Fabre et al.
Langmuir : the ACS journal of surfaces and colloids, 28(5), 2835-2841 (2012-01-11)
Skeletonized zirconium phosphonate surfaces are used to support planar lipid bilayers and are shown to be viable substrates for studying transmembrane proteins. The skeletonized surfaces provide space between the bilayer and the solid support to enable protein insertion and avoid
Sajjad M Waraich et al.
Journal of colloid and interface science, 367(1), 74-82 (2011-11-22)
Composite materials with an ordered layered structure resembling that of nacre were fabricated by layer-by-layer assembly making use of presynthesized α-zirconium hydrogenphosphate hydrate (ZrP) platelets and chitosan. These two biocompatible materials were chosen in view of possible applications in the
Ajith Pattammattel et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 2971-2981 (2013-02-05)
Previously, an ion-coupled protein binding (ICPB) model was proposed to explain the thermodynamics of protein binding to negatively charged α-Zr(IV) phosphate (α-ZrP). This model is tested here using glucose oxidase (GO) and met-hemoglobin (Hb) and several cations (Zr(IV), Cr(III), Au(III)
Weijie Qin et al.
Analytical chemistry, 82(22), 9461-9468 (2010-10-30)
Reversible phosphorylation is one of the most important post-translational modifications of proteins and a key regulator of cellular signaling pathways. Specific enrichment of phosphopeptides from proteolytic digests is a prerequisite for large scale identification of protein phosphorylation by mass spectrometry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service