Skip to Content
Merck
All Photos(2)

Key Documents

449628

Sigma-Aldrich

Aluminum fluoride

anhydrous, powder, 99.8% trace metals basis

Synonym(s):

Aluminum trifluoride, Trifluoroaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlF3
CAS Number:
Molecular Weight:
83.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.8% trace metals basis

form

powder

impurities

≤1,000.0 ppm Trace Metal Analysis

mp

1290 °C (lit.)

density

3.1 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

F[Al](F)F

InChI

1S/Al.3FH/h;3*1H/q+3;;;/p-3

InChI key

KLZUFWVZNOTSEM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lecoq, A.; et al.
Non-Cryst. Solids, 41, 209-209 (1980)
Applied Physics Letters, 69, 2012-2012 (1996)
Sun, Y.-K.; et al.
Electrochemical Communications, 8, 821-821 (2006)
Cheng-Chung Lee et al.
Applied optics, 44(34), 7333-7338 (2005-12-16)
Aluminum fluoride (AlF3) was deposited by a resistive heating boat. To obtain a low optical loss and high laser-induced damage threshold (LIDT) at 193 nm, the films were investigated under different substrate temperatures, deposition rates, and annealing after coating. The
Electronics Letters, 32, 1586-1586 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service