Skip to Content
Merck
All Photos(1)

Key Documents

463221

Sigma-Aldrich

Glycolic acid ethoxylate lauryl ether

average Mn ~360

Synonym(s):

Laureth-4 carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11-13(OCH2CH2)nOCH2CO2H
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

mol wt

average Mn ~360

refractive index

n20/D 1.45

density

1 g/mL at 25 °C (lit.)

SMILES string

OCCO.OCC(O)=O.CCCCCCCCCCCCO.CCCCCCCCCCCCCO.CCCCCCCCCCCCCCO

InChI

1S/C14H30O.C13H28O.C12H26O.C2H4O3.C2H6O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15;1-2-3-4-5-6-7-8-9-10-11-12-13-14;1-2-3-4-5-6-7-8-9-10-11-12-13;3-1-2(4)5;3-1-2-4/h15H,2-14H2,1H3;14H,2-13H2,1H3;13H,2-12H2,1H3;3H,1H2,(H,4,5);3-4H,1-2H2

InChI key

KQSLVJFTAGAPJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Surfactant.
Stabilizer for microemulsions.

Features and Benefits

Not sensitive to hard water.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service