Skip to Content
Merck
All Photos(3)

Documents

STS0212

Sigma-Aldrich

ECO BRIJ® L4

greener alternative

average Mn ~362

Synonym(s):

Brij® L4, Polyethylene glycol dodecyl ether, Polyoxyethylene (4) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H42O5)n
CAS Number:
MDL number:
UNSPSC Code:
12165104
NACRES:
NA.28

description

non-ionic

Quality Level

form

semisolid (may be liquid)

mol wt

average Mn ~362

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1.0% water

refractive index

n20/D 1.451 (lit.)

mp

48 °C

acid number

≤2.0 mg KOH/g

hydroxyl value

144‑165 mg KOH/g

solubility

water: insoluble

density

0.95 g/mL at 25 °C (lit.)

HLB

9.7

greener alternative category

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCO

InChI

1S/C20H42O5/c1-2-3-4-5-6-7-8-9-10-11-13-22-15-17-24-19-20-25-18-16-23-14-12-21/h21H,2-20H2,1H3

InChI key

WPMWEFXCIYCJSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ECO Brij® L4, also known as polyethylene glycol lauryl ether, is a bio-based, nonionic surfactant manufactured from naturally occurring straight-chain lauryl alcohol. This detergent, derived from natural sources, provides various functional advantages, such as detergency, emulsification, and wetting, making it suitable for a range of applications in biochemical and biological research.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions
  • High-purity chemical suitable for a wide variety of research applications

Physical properties

The ECO Brij series of ethoxylated fatty alcohols are non-ionic surfactants manufactured using renewable sources. ECO Brij L4 is a mixture of tetraethylene glycol alkyl ethers, primarily dodecyl (C12) ether. The alkyl ether profile of ECO Brij L4 may differ lot-to-lot due to natural variability in the lipid starting material (palm oil).

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Brij is a registered trademark of Croda International PLC
ECO BRIJ is a registered trademark of Croda Inc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina G Fabi et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 38(7 Pt 2), 1112-1127 (2012-01-25)
The aged hand is characterized by cutaneous and dermal atrophy, with deep intermetacarpal spaces, prominent bones and tendons, and bulging reticular veins. Epidermal changes include solar lentigines, seborrheic keratoses, actinic keratoses, skin laxity, rhytides, tactile roughness, and telangiectasia. A Medline
Adriana Smarandache
Photomedicine and laser surgery, 30(5), 262-267 (2012-03-16)
The goal of this study was to investigate the laser light scattering involved in the laser irradiation process of polidocanol foam samples. Clinical experimental results proved that exposure of tissues impregnated with foaming polidocanol to laser radiation emitted at 1064
S Reich-Schupke et al.
VASA. Zeitschrift fur Gefasskrankheiten, 41(4), 243-247 (2012-07-25)
According to the guidelines and the manufacturer's information, pregnancy is a contraindication for sclerotherapy with Polidocanol. However, in some cases sclerotherapy has been conducted in a period when the pregnancy is not known by the patient. When pregnancy is diagnosed
J-M Gonzalez et al.
Endoscopy, 44(5), 539-542 (2012-03-06)
Recent developments in therapeutic endoscopic ultrasound (EUS) have enabled new approaches to the management of refractory gastrointestinal bleeding, including EUS-guided sclerotherapy and vessel embolization. Few cases have been reported in the literature. Eight patients were admitted for severe, refractory gastrointestinal
Albert-Adrien Ramelet
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 38(5), 748-751 (2012-01-25)
Some leg telangiectasias may be refractory to treatment, including sclerotherapy and lasers. To describe the innovative Sclerotherapy in Tumescent Anesthesia of Reticular veins and Telangiectasias (START) approach to achieving good results in such patients, which also proves effective in treating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service