Skip to Content
Merck
All Photos(1)

Key Documents

449601

Sigma-Aldrich

Aluminum bromide

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Aluminum tribromide, Aluminum tribromide (AlBr3 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlBr3
CAS Number:
Molecular Weight:
266.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 81.3 °C)

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: aluminum

impurities

≤15.0 ppm Trace Metal Analysis

mp

94-98 °C (lit.)

density

3.205 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Br[Al](Br)Br

InChI

1S/Al.3BrH/h;3*1H/q+3;;;/p-3

InChI key

PQLAYKMGZDUDLQ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service