Skip to Content
Merck
All Photos(1)

Key Documents

444014

Sigma-Aldrich

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol

97%

Synonym(s):

1,1,1,3,3,3-Hexafluoro-2-hydroxy-2-methylpropane, 1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, 1,1,1-Trifluoro-2-trifluoromethyl-2-propanol, 1,1-Bis(trifluoromethyl)ethanol, 2-Methylhexafluoro-2-propanol, Hexafluoro-tert-butanol, Hexafluoro-tert-butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(CF3)2OH
CAS Number:
Molecular Weight:
182.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.3 (lit.)

bp

60-62 °C (lit.)

density

1.484 g/mL at 25 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

CC(O)(C(F)(F)F)C(F)(F)F

InChI

1S/C4H4F6O/c1-2(11,3(5,6)7)4(8,9)10/h11H,1H3

InChI key

FQDXJYBXPOMIBX-UHFFFAOYSA-N

Application

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol can be used as a reactant to synthesize:
  • Ethyl zinc fluoroalkoxide complexes by using diethyl zinc.
  • Vanadium-alkylidene complexes in the presence of hexane.
  • 1,1,1,3,3,3-Hexafluoro-2-methyl-2-propylperchlorate using NaH and dichlorine heptoxide in carbon tetrachloride.
It can also be used as a solvent to prepare cyclodextrin-assembled material.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of dichlorine heptoxide with alcohols
Baum, K, et al.
Journal of the American Chemical Society, 96, 3233-3237 (1974)
Toshiyuki Kida et al.
Chemical communications (Cambridge, England), 50(91), 14245-14248 (2014-10-07)
We found that cyclodextrins (CDs) have a high solubility in 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP). Evaporating HFIP from CD solutions on a glass plate gave crystalline solids composed of channel-type CD assemblies, and electrospinning with an HFIP solution of CDs fabricated CD microfibers.
Synthesis of ethyl zinc fluoroalkoxide complexes and the crystal structure of [EtZn (py){?-OCH (CF3)2}]2
Suh S, et al.
Polyhedron, 27, 513-516 (2008)
Inorganic Chemistry, 33, 5463-5463 (1994)
Chemistry of Materials, 7, 929-929 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service