Skip to Content
Merck
All Photos(1)

Key Documents

538795

Sigma-Aldrich

2-Trifluoromethyl-2-propanol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C(CH3)2OH
CAS Number:
Molecular Weight:
128.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.3335 (lit.)

bp

83 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

CC(C)(O)C(F)(F)F

InChI

1S/C4H7F3O/c1-3(2,8)4(5,6)7/h8H,1-2H3

InChI key

OCGWWLDZAFOHGD-UHFFFAOYSA-N

General description

2-Trifluoromethyl-2-propanol is a fluorinated aliphatic alcohol. It can be prepared by reacting methyllithium, trifluoroacetone and trifluorovinyl bromide in ether. This can also be synthesized from the reaction between methylmagnesium bromide and 1,1,1-trifluoroacetone in ether.

Application

2-Trifluoromethyl-2-propanol may be employed as a solvent for the preparation of [18F]fluorothymidine. It may also be used in the synthesis of [2-(trifluoromethyl)-2-propyl nitrate] through nitration in nitric acid/trifluoroacetic anhydride.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanistic studies on gas-phase negative ion unimolecular decompositions. Alkoxide anions
Tumas W, et al.
Journal of the American Chemical Society, 110.9 , 2714-2722 (1988)
The Preparation and Some Reactions of Trifluorovinyllithium1
Tarrant P, et al.
The Journal of Organic Chemistry, 28.3, 839-843 (1963)
Nonacidic nitration of secondary amines
Bottaro JC, et al.
The Journal of Organic Chemistry, 52.11 , 2292-2294 (1987)
Comparison of synthesis yields of 3'-deoxy-3'-[18F] fluorothymidine by nucleophilic fluorination in various alcohol solvents
Lee SJ, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 51.1, 80-82 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service