Skip to Content
Merck
All Photos(1)

Documents

257478

Sigma-Aldrich

2,2,3,3,3-Pentafluoro-1-propanol

97%

Synonym(s):

PFPOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CH2OH
CAS Number:
Molecular Weight:
150.05
Beilstein:
1743133
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.288 (lit.)

bp

80 °C/748 mmHg (lit.)

density

1.505 g/mL at 25 °C (lit.)

SMILES string

OCC(F)(F)C(F)(F)F

InChI

1S/C3H3F5O/c4-2(5,1-9)3(6,7)8/h9H,1H2

InChI key

PSQZJKGXDGNDFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,3,3,3-Pentafluoro-1-propanol is an alternative cleaning agent for chlorofluorocarbon.

Application

2,2,3,3,3-Pentafluoro-1-propanol has been used:
  • as derivatization reagent in detection of unlabeled and 15N2 -labeled L-tryptophan, L-kynurenine, serotonin and quinolinic acid in human and rat plasma by GC-MS
  • as derivatization reagent in simultaneous analysis of cocaine, cocaethylene and their possible metabolic and pyrolytic products in blood, urine and muscle by GC-MS
  • preparation of trifluoromethyl ynamines which, in turn, converted aldehydes to α-trifluoromethyl-α,β-unsaturated amides
  • to generate fluorinated α-keto ethers with alkenes and has wide applications in the expanding fluorous research area

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Liver

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ishihara, T.; et al.
Tetrahedron, 62, 3783-3783 (2006)
M Usami et al.
Kokuritsu Iyakuhin Shokuhin Eisei Kenkyujo hokoku = Bulletin of National Institute of Health Sciences, (118)(118), 45-49 (2001-09-06)
Teratogenicity of 2,2,3,3,3-pentafluoro-1-propanol (5FP), an alternative cleaning agent for chlorofluorocarbon, was examined in rats. 5FP was diluted with sesame oil and given to pregnant rats (Crj: Wistar) by gavage once a day from day 7 to 17 of pregnancy at
Mitsue Sano et al.
Journal of mass spectrometry : JMS, 49(2), 128-135 (2014-03-29)
A method for the detection of unlabeled and (15)N2 -labeled L-tryptophan (L-Trp), L-kynurenine (L-Kyn), serotonin (5-HT) and quinolinic acid (QA) in human and rat plasma by GC/MS is described. Labeled and unlabeled versions of these four products were analyzed as
Patrick S Cardona et al.
Forensic science international, 157(1), 46-56 (2006-01-24)
A method was developed for simultaneously analyzing cocaine (COC), benzoylecgonine (BZE), norbenzoylecgonine (BNE), norcocaine (NCOC), ecgonine (ECG), ecgonine methyl ester (EME), m-hydroxybenzoylecgonine (HBZE), anhydroecgonine methyl ester (AEME), cocaethylene (CE), norcocaethylene (NCE), and ecgonine ethyl ester (EEE) in blood, urine, and
Sudha Manandhar et al.
The Journal of organic chemistry, 67(18), 6415-6420 (2002-08-31)
Interactions of various fluorinated and nonfluorinated alcohols with trans-stilbene in the presence of electrophilic reagents were studied. Under neat conditions, reactions of trans-stilbene (1) with fluorinated alcohols, R(f)OH (R(f) = CF3CH2-, CFH2CH2-, CF3CF2CH2-, CF2H(CF2)3CH2-, (CF3)2CH-, (CF3)3C- (2a-f) in the presence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service