Skip to Content
Merck
All Photos(1)

Key Documents

441619

Sigma-Aldrich

(1S)-(+)-3-Carene

99%

Synonym(s):

(+)-3-Carene, (1S)-3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1902767
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

optical activity

[α]20/D +17°, neat

refractive index

n20/D 1.472 (lit.)

bp

170-172 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

[H][C@@]12CC=C(C)C[C@]1([H])C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-9(6-7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9+/m1/s1

InChI key

BQOFWKZOCNGFEC-BDAKNGLRSA-N

General description

(1S)-(+)-3-Carene is a monoterpene.[1]

Application

(1S)-(+)-3-Carene has been used as a standard during the quantification of components in Lavandula stoechas essential oils by fast gas chromatography-mass spectrometry (FGC-EI/MS).[2] It may be used as a starting material in the total synthesis of (+)-ingenol.[3] It may also be used to synthesize (1S,3S,4S,6R)-3,7,7-trimethyl-4-morpholino-4-yl-bicyclo(4.1.0)heptano-3-ol, which can catalyze the addition of diethylzinc to different aldehydes with high enantioselectivity.[4]
Chiral building block for the preparation of bicyclo[3.2.0]heptenes that are useful intermediates in the synthesis of (+)-lineatin and both enantiomers of capnellene.[5] Has been converted to its corresponding allenyl allylic ether[6] for studies of its thermal isomerization chemistry. Precursor to di(4-isocaranyl)borane, an asymmetric hydroboration reagent for olefins.[7]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective addition of diethylzinc to aldehydes catalyzed by a ?-amino alcohol derived from (+)-3-carene.
Joshi SN and Malhotra SV.
Tetrahedron Asymmetry, 14(13), 1763-1766 (2003)
Xiaofeng Zhu et al.
Environmental entomology, 49(3), 586-592 (2020-03-22)
Scolytus schevyrewi Semenov (Coleoptera: Curculionidae: Scolytinae) is native to China and Central Asia. Damage by the adults and larvae weakens local apricot trees (Armeniaca spp.), often causing death to many host trees. In previous studies, freshly cut apricot logs were
Dulcere, J.-P. et al.
The Journal of Organic Chemistry, 58, 5702-5702 (1993)
Sonawane, H.R. et al.
Synlett, 875-875 (1993)
Brown, H.C. et al.
The Journal of Organic Chemistry, 53, 2911-2911 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service