Skip to Content
Merck
All Photos(2)

Key Documents

115576

Sigma-Aldrich

3-Carene

90%

Synonym(s):

δ3-Carene, 3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

optical activity

[α]20/D +15°, neat

impurities

5% 2-carene

refractive index

n20/D 1.474 (lit.)

bp

168-169 °C/705 mmHg (lit.)

density

0.857 g/mL at 25 °C (lit.)

SMILES string

CC1=CCC2C(C1)C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-9(6-7)10(8,2)3/h4,8-9H,5-6H2,1-3H3

InChI key

BQOFWKZOCNGFEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smail Aazza et al.
Molecules (Basel, Switzerland), 16(9), 7672-7690 (2011-09-09)
The commercial essential oils of Citrus aurantium L., Cupressus sempervirens L., Eucalyptus globulus Labill., Foeniculum vulgare Mill. and Thymus vulgaris L., isolated by steam distillation by a company of Morocco were evaluated in terms of in vitro antioxidant activity through
Asa M Jonsson et al.
Environmental science & technology, 40(1), 188-194 (2006-01-26)
The effect of water on the initial secondary organic aerosol (SOA) formation from gas-phase ozonolysis of limonene, delta3-carene, and alpha-pinene (-3 and approximately 1.5 x 10(11) molecule cm(-3) reacted) has been investigated in a flow reactor at controlled relative humidity
Dawn E Hall et al.
The Plant journal : for cell and molecular biology, 65(6), 936-948 (2011-02-18)
Conifers are extremely long-lived plants that have evolved complex chemical defenses in the form of oleoresin terpenoids to resist attack from pathogens and herbivores. In these species, terpenoid diversity is determined by the size and composition of the terpene synthase
Yan Ma et al.
Physical chemistry chemical physics : PCCP, 11(21), 4184-4197 (2009-05-22)
This paper describes experimental studies aimed at elucidating mechanisms for the formation of low-volatility organic acids in the gas-phase ozonolysis of 3-carene. Experiments were carried out in a static chamber under 'OH-free' conditions. A range of multifunctional acids-which are analogous
Min Lu et al.
Ecology, 92(11), 2013-2019 (2011-12-15)
Novel genotypes often arise during biological invasions, but their role in invasion success has rarely been elucidated. Here we examined the population genetics and behavior of the fungus, Leptographium procerum, vectored by a highly invasive bark beetle, Dendroctonus valens, to

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service