Skip to Content
Merck
All Photos(1)

Documents

441562

Sigma-Aldrich

trans-1-Acetyl-4-hydroxy-L-proline

≥98%, for peptide synthesis

Synonym(s):

N-Acetyl-L-hydroxyproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO4
CAS Number:
Molecular Weight:
173.17
Beilstein:
84043
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

product name

trans-1-Acetyl-4-hydroxy-L-proline, ≥98%

Assay

≥98%

form

powder

optical activity

[α]20/D −119°, c = 4 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

132-133 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC(=O)N1C[C@H](O)C[C@H]1C(O)=O

InChI

1S/C7H11NO4/c1-4(9)8-3-5(10)2-6(8)7(11)12/h5-6,10H,2-3H2,1H3,(H,11,12)/t5-,6+/m1/s1

InChI key

BAPRUDZDYCKSOQ-RITPCOANSA-N

Application

trans-1-Acetyl-4-hydroxy-L-proline can be used:
  • In the stereospecific synthesis of 4-fluoroglutamic acid.
  • To synthesize molecular targets for von Hippel-Lindau (VHL) E3 ubiquitin ligase.
  • As a precursor to synthesize pseudopoly(amino acids) such as poly(trans-4-hydroxy-4-acyl-L-proline ester) and a biodegradable polymer, poly(lactic acid-glycolic acid-4-hydroxyproline).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure-guided design and optimization of small molecules targeting the protein?protein interaction between the von Hippel?Lindau (VHL) E3 ubiquitin ligase and the hypoxia inducible factor (HIF) alpha subunit with in vitro nanomolar affinities.
Galdeano C, et al.
Journal of Medicinal Chemistry, 57(20), 8657-8663 (2014)
Pseudopoly (amino acids): A Study of the Synthesis and Characterization of Poly (trans-4-hydroxy-N-acyl-L-proline esters).
Kwon H Y and Langer R
Macromolecules, 22(8), 3250-3255 (1989)
Synthesis and characterization of a novel biodegradable polymer poly (lactic acid?glycolic acid?4?hydroxyproline).
Duan J, et al.
Journal of Applied Polymer Science, 103(6), 3585-3590 (2007)
P Villani et al.
Presse medicale (Paris, France : 1983), 27(5), 211-214 (1998-10-13)
Assess the importance of the mid-term placebo effect of symptomatic slow acting drugs given for osteoarthritis. We analyzed six controlled trials available in the literature. Trial duration ranged from 2 to 6 months. The trials had been conducted to assess
A Harris et al.
European journal of medical research, 3(4), 182-188 (1998-05-16)
Oxaceprol, an established drug for treatment of degenerative joint disease, has recently been shown in vitro to reduce leukocyte adhesion to cultured endothelial cells and leukocyte extravasation in vivo in an arthritis animal model. The aim of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service