Skip to Content
Merck
All Photos(1)

Key Documents

436240

Sigma-Aldrich

BRIJ® O20

average Mn ~1,150

Synonym(s):

Polyoxyethylene (20) oleyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H35(OCH2CH2)nOH, n~20
CAS Number:
Molecular Weight:
1149.53
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

non-ionic

Quality Level

mol wt

average Mn ~1,150

technique(s)

cell culture | mammalian: suitable
protein quantification: suitable

mp

25-30 °C (lit.)

HLB

15

SMILES string

O(CCO)CCCCCCCC\C=C/CCCCCCCC

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h9-10,21H,2-8,11-20H2,1H3/b10-9-

InChI key

KWVPFECTOKLOBL-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brij® O20 is a non-ionic surfactant that has an 18 carbon tail with a single double bond and the head contains 20 ethylene oxide moieties. It is polyethoxylated fatty alcohol that is hydrophilic in nature.

Application

Brij® O20 has been used:
  • as a detergent in medium A to prepare single detergent micelle aggregates
  • along with phosphate buffer saline (PBS) solution as the receptor solution for terbinafine (TBF) for in vitro permeation studies of TBF-free base in porcine skin
  • as a detergent to isolate detergent-resistant membranes from the mouse brain and to determine the distribution of major mammalian brain gangliosides, GM1, GD1a, GD1b and GT1b

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>464.0 °F - Equilibrium method

Flash Point(C)

> 240 °C - Equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mixed Micellization of Polyoxyethylene (20) Oleyl Ether with Cetylpyridinium Chloride at the Air-Water Interface
Desai S and Bhagwat SS
Journal of Surfactants and Detergents, 19(6), 1169-1174 (2016)
Pulkit Khatri et al.
Journal of pharmaceutical sciences, 107(8), 2198-2207 (2018-04-08)
The study aims to elucidate the mechanism and the role of the molecular structure of surfactants and lipids in the formation of oil-in-water (o/w) self-emulsified nanoemulsions (SENs). The hypothesis is that the overall change of Gibb's free energy (ΔGf) during
Towards a new universal way to describe the required hydrophilic lipophilic balance of oils using the phase inversion temperature of C10E4/n-octane/water emulsions
Royer M, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 536, 165-171 (2018)
Gunasekaran Dhandapani et al.
Biotechnology and bioengineering, 119(7), 1997-2003 (2022-03-25)
Immunoglobulin M (IgM) antibodies hold promise as anticancer drugs and as agents for promoting immune homeostasis. This promise has not been realized due to low expression levels in mammalian cells producing IgM class antibodies, and the failure of protein A
María Vergara-Barberán et al.
Journal of agricultural and food chemistry, 59(20), 10775-10780 (2011-09-13)
A method for the determination of fatty acids in vegetable oils by capillary electrophoresis with indirect UV-vis detection has been developed. The separation of fatty acids was optimized in terms of Brij surfactant nature and concentration and organic modifier (2-propanol)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service