Skip to Content
Merck
All Photos(3)

Key Documents

466387

Sigma-Aldrich

Brij® S 100

average Mn ~4,670

Synonym(s):

Polyoxyethylene (100) stearyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H37(OCH2CH2)nOH, n~100
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~4,670

Quality Level

mp

51-54 °C (lit.)

hydroxyl value

13‑25 mg KOH/g

solubility

propylene glycol and xylene: insoluble

density

1.1 g/mL at 25 °C (lit.)

HLB

18

SMILES string

O(CC[O])CCCCCCCCCCCCCCCCCC

InChI

1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3

InChI key

PSEGVHKUPXNGGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Brij® S 100 may be used:
  • as a non-ionic surfactant to study its interactions with luminol electrochemiluminescence for bioanalysis
  • as a carrier material to study the formulation of nimodipine, fenofibrate, and o-vanillin using the particles from gas-saturated solution (PGSS) process
  • as a hydrophilic surfactant in the casting solution to prepare an asymmetric flat sheet nanofiltration membranes and to study its effects on the hydrophilic property of the poly(ethersulfone) (PES) nanofiltration membranes

Legal Information

Brij is a registered trademark of Croda International PLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Formulation of nimodipine, fenofibrate, and o-vanillin with Brij S100 and PEG 4000 using the PGSS process
Kravanja G, et al.
Journal of Supercritical Fluids, 135, 245-253 (2018)
J Q Zhang et al.
Drug delivery, 14(6), 381-387 (2007-08-19)
The study describes the development of stealth solid lipid nanoparticles (SLNs) as colloidal carriers for silibinin, a drug with very low solubility. Stealth SLNs were constituted mainly of bioacceptable and biodegradable lipids, such as stearic acid and surfactant Brij 78
S O Williams et al.
Drug development and industrial pharmacy, 26(1), 71-77 (2000-04-01)
The purpose of this study was to scale up an oil/water (o/w) cream formulation containing 40% diethylene glycol monoethyl ether (DGME), developed via 300-g laboratory batches in a 2(5-2) fractional factorial design, to 7-kg batch sizes in a Brogli-10 homogenizer.
Jae-Haeng Song et al.
Drug development and industrial pharmacy, 35(8), 897-903 (2009-02-26)
The oral administration of loratadine, an antihistamine, can have a variety of adverse side effects, such as headache, fatigue, and nausea, because of the transient high blood concentration. To avoid these effects, loratadine can be administered using a transdermal drug
Y C Lee et al.
Journal of pharmaceutical sciences, 86(4), 430-433 (1997-04-01)
An ocular insert is developed for the controlled systemic delivery of insulin. Commercially available Gelfoam absorbable gelatin sponge, USP, is used in the fabrication of the ocular insert in the form of a matrix system. Two eyedrop formulations and 13

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service