Merck
  • Ultrasonic-assisted cloud point extraction for determination of nickel in water samples by flame atomic absorption spectrometry.
Ultrasonic-assisted cloud point extraction for determination of nickel in water samples by flame atomic absorption spectrometry.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-07-07)
Jun Song, Wei Zhen, Zonghao Li, Yuanpei Lian, Yaling Yang
ABSTRACT
A novel method for the determination of nickel was established by ultrasonic-assisted cloud point extraction (UA-CPE) prior to flame atomic absorption spectrometry (FAAS) analysis. The nickel reacted with N,N'-bis(salicylidene)-1,2-ethanediamine (BSE) to form hydrophobic chelates, which were extracted into the micelles of alpha-[3,5-dimethyl-1-(2-methylpropyl)hexyl]-omega- poly(oxy-2-ethanediyl) (Tergitol TMN-6). Tergitol TMN-6 was used as green nonionic surfactant. BSE was synthesized and checked by nuclear magnetic resonance (NMR) spectra. The phase diagrams of the binary system, water-surfactant (Tergitol TMN-6), and the ternary systems, water-surfactant-salt, were determined. The effects of experimental conditions including pH of sample solution, concentration of chelating agent and surfactant, ultrasonic power, equilibration temperature and incubation time were evaluated in order to enhance sensitivity of the method. Under the optimal conditions, the calibration graph was linear in the range of 10-500 μg L(-1). The values obtained for the limit of detection and enrichment factor were 1.0 μg L(-1) and 30, respectively. The method was successfully applied to the analysis of nickel in water samples.
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
TERGITOL, Type NP-9
Sigma-Aldrich
TERGITOL solution, Type NP-40, 70% in H2O
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800